MilliporeSigma
  • Remote ischemic post-conditioning improves neurological function by AQP4 down-regulation in astrocytes.
Remote ischemic post-conditioning improves neurological function by AQP4 down-regulation in astrocytes.
Behavioural brain research (2015-04-25)
Shuai Li, Xiaosong Hu, Mingxiao Zhang, Fangfang Zhou, Na Lin, Qingjie Xia, Yu Zhou, Wenqian Qi, Yonghua Zong, Huijun Yang, Tinghua Wang
ABSTRACT
Cerebral ischemia is a primary cause of human death and long-term disability. Previous studies have suggested that remote ischemic post-conditioning (RIPC) is a potential useful tool for cerebral ischemic treatment. However, the protective mechanism of RIPC is not very clear. This study verified the hypothesis that, in remote post-conditioning of cerebral ischemic rats, down-regulation of aquaporin 4 (AQP4), which is an important player for water hemostasis in astrocytes, could attenuate cerebral damage after transient middle cerebral artery occlusion (MCAO). In this study, RIPC model was established after MCAO. Each hind limb of rat was clamped by small rubber tubes for 10 min, and then the tubes were opened for 10 min. The clamping and opening were operated for a total of three cycles to block the hind limbs blood flow. The results showed that, RIPC could significantly improve neurological function, decrease the percentage of the infarct volume and edema, and elevate the integrity of blood-brain barrier (BBB). In addition, the numbers of AQP4 and glial fibrillary acidic protein (GFAP) positive cells were significantly lower in the RIPC group. Moreover, we found that AQP4 expression decreased in response to ischemia/reperfusion in the RIPC group. Our findings indicated that RIPC could attenuate focal cerebral ischemia/reperfusion injury, and the neuroprotective mechanism was related with the down-regulation of AQP4 in astrocytes.
Sigma-Aldrich
2,3,5-Triphenyltetrazolium chloride, JIS special grade, 98.0-102.0%
Sigma-Aldrich
Hydrogen peroxide solution, SAJ first grade, ≥30.0%
Sigma-Aldrich
Hydrogen peroxide solution, contains potassium stannate as inhibitor, 30-32 wt. % in water, semiconductor grade, 99.999% trace metals basis
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
2,3,5-Triphenyltetrazolium chloride, ≥98.0% (HPLC)
Sigma-Aldrich
Formamide, SAJ first grade, ≥98.5%
Sigma-Aldrich
Formamide, ≥99.5% (GC), BioReagent, Molecular Biology
Sigma-Aldrich
Formamide, BioUltra, Molecular Biology, ≥99.5% (T)
Sigma-Aldrich
Formamide, JIS special grade, ≥98.5%
Sigma-Aldrich
Formamide, ACS reagent, ≥99.5%
Sigma-Aldrich
Formamide solution, suitable for NMR (reference standard), 90% in DMSO-d6 (99.9 atom % D), NMR tube size 10 mm × 8 in.
Supelco
Formamide solution, suitable for NMR (reference standard), 90% in DMSO-d6 (99.9 atom % D), NMR tube size 5 mm × 8 in.
Sigma-Aldrich
DAPI, for nucleic acid staining