MilliporeSigma
Synthesis of the metabolite N-hydroxy-desferrioxamine B.
The Journal of antibiotics (1993-02-01)
B E Roggo, H H Peter
ABSTRACT
N-Hydroxy-desferrioxamine B (5), a postulated metabolite of the microbial product desferrioxamine B (1), has been prepared by reduction of the intermediate oxime 6 with sodium cyanoborohydride. The oxime was obtained by selective oxidation of desferrioxamine B with hydrogen peroxide and a catalytic amount of sodium tungstate dihydrate. The iron complex derived from 5 enabled definite proof of the structure of one of four metabolites of desferrioxamine B found in urine samples of patients treated with this drug.
Sigma-Aldrich
Sodium tungstate dihydrate, ACS reagent, ≥99%
Pricing and availability is not currently available.