MilliporeSigma
A transmembrane CXC chemokine is a ligand for HIV-coreceptor Bonzo.
Nature immunology (2001-03-23)
M Matloubian, A David, S Engel, J E Ryan, J G Cyster
ABSTRACT
We describe a protein with the hallmarks of a chemokine, designated CXCL16, that is made by dendritic cells (DCs) in lymphoid organ T cell zones and by cells in the splenic red pulp. CXCL16 contains a transmembrane domain and both membrane-bound and soluble forms are produced. Naïve CD8 T cells, natural killer T cells and a subset of memory CD4 T cells bind CXCL16, and activated T cells migrated chemotactically to the soluble chemokine. By expression cloning, Bonzo (also known as STRL33 and TYMSTR) was identified as a CXCL16 receptor. CXCL16 may function in promoting interactions between DCs and CD8 T cells and in guiding T cell movements in the splenic red pulp. CXCL16 was also found in the thymic medulla and in some nonlymphoid tissues, indicating roles in thymocyte development and effector T cell trafficking.
Sigma-Aldrich
CXCL16 from mouse, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture
Pricing and availability is not currently available.
Sigma-Aldrich
CXCL16 human, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture
Pricing and availability is not currently available.