MilliporeSigma
  • Structure-activity relationships among antifungal nylon-3 polymers: identification of materials active against drug-resistant strains of Candida albicans.
Structure-activity relationships among antifungal nylon-3 polymers: identification of materials active against drug-resistant strains of Candida albicans.
Journal of the American Chemical Society (2014-03-13)
Runhui Liu, Xinyu Chen, Shaun P Falk, Brendan P Mowery, Amy J Karlsson, Bernard Weisblum, Sean P Palecek, Kristyn S Masters, Samuel H Gellman
ABSTRACT
Fungal infections are a major challenge to human health that is heightened by pathogen resistance to current therapeutic agents. Previously, we were inspired by host-defense peptides to develop nylon-3 polymers (poly-β-peptides) that are toxic toward the fungal pathogen Candida albicans but exert little effect on mammalian cells. Based on subsequent analysis of structure-activity relationships among antifungal nylon-3 polymers, we have now identified readily prepared cationic homopolymers active against strains of C. albicans that are resistant to the antifungal drugs fluconazole and amphotericin B. These nylon-3 polymers are nonhemolytic. In addition, we have identified cationic-hydrophobic copolymers that are highly active against a second fungal pathogen, Cryptococcus neoformans, and moderately active against a third pathogen, Aspergillus fumigatus.
Product Description
Millipore
YPD Broth, NutriSelect® Basic, powder, for fungi, poly bottle of 250 g, poly bottle of 1 kg
Pricing and availability is not currently available.