MilliporeSigma
  • Determination of coenzyme Q10 content in raw materials and dietary supplements by high-performance liquid chromatography-UV: collaborative study.
Determination of coenzyme Q10 content in raw materials and dietary supplements by high-performance liquid chromatography-UV: collaborative study.
Journal of AOAC International (2008-08-30)
Steven Lunetta, Mark Roman
ABSTRACT
An international collaborative study was conducted of a high-performance liquid chromatographic (HPLC)-UV method for the determination of coenzyme Q10 (CoQ10, ubidecarenone) in raw materials and dietary supplements. Ten collaborating laboratories determined the total CoQ10 content in 8 blind duplicate samples. Sample materials included CoQ10 raw material and 4 finished product dietary supplements representing softgels, hardshell gelatin capsules, and chewable wafers. In addition, collaborating laboratories received a negative control and negative control spiked with CoQ10 at low and high levels to determine recovery. Materials were extracted with an acetonitrile-tetrahydrofuran-water mixture. Ferric chloride was added to the test solutions to ensure all CoQ10 was in the oxidized form. The HPLC analyses were performed on a C18 column using UV detection at 275 nm. Repeatability relative standard deviations (RSDr) ranged from 0.94 to 5.05%. Reproducibility relative standard deviations (RSDR) ranged from 3.08 to 17.1%, with HorRat values ranging from 1.26 to 5.17. Recoveries ranged from 74.0 to 115%. Based on these results, the method is recommended for Official First Action for determination of CoQ10 in raw materials and dietary supplement finished products containing CoQ10 at a concentration of >100 mg CoQ10/g test material.
Sigma-Aldrich
Coenzyme Q10, ≥98% (HPLC)
Pricing and availability is not currently available.