MilliporeSigma
  • Glyoxal and methylglyoxal as urinary markers of diabetes. Determination using a dispersive liquid-liquid microextraction procedure combined with gas chromatography-mass spectrometry.
Glyoxal and methylglyoxal as urinary markers of diabetes. Determination using a dispersive liquid-liquid microextraction procedure combined with gas chromatography-mass spectrometry.
Journal of chromatography. A (2017-06-24)
M Pastor-Belda, A J Fernández-García, N Campillo, M D Pérez-Cárceles, M Motas, M Hernández-Córdoba, P Viñas
ABSTRACT
Glyoxal (GO) and methylglyoxal (MGO) are α-oxoaldehydes that can be used as urinary diabetes markers. In this study, their levels were measured using a sample preparation procedure based on salting-out assisted liquid-liquid extraction (SALLE) and dispersive liquid-liquid microextraction (DLLME) combined with gas chromatography-mass spectrometry (GC-MS). The effect of the derivatization reaction with 2,3-diaminonaphthalene, the addition of acetonitrile and sodium chloride to urine, and the DLLME step using the acetonitrile extract as dispersant solvent and carbon tetrachloride as extractant solvent were carefully optimized. Quantification was performed by the internal standard method, using 5-bromo-2-chloroanisole. The intraday and interday precisions were lower than 6%. Limits of detection were 0.12 and 0.06ngmL
Sigma-Aldrich
5-Bromo-2-chloroanisole, 97%
Pricing and availability is not currently available.
Sigma-Aldrich
Creatinine, anhydrous, ≥98%
Pricing and availability is not currently available.