MilliporeSigma
  • Antimicrobial peptide LL-37 promotes the viability and invasion of skin squamous cell carcinoma by upregulating YB-1.
Antimicrobial peptide LL-37 promotes the viability and invasion of skin squamous cell carcinoma by upregulating YB-1.
Experimental and therapeutic medicine (2017-07-05)
Wei Wang, Yan Zheng, Jinjing Jia, Changji Li, Qiqi Duan, Ruilian Li, Xin Wang, Yongping Shao, Caifeng Chen, Huling Yan
ABSTRACT
Antimicrobial peptide LL-37 serves a function in the host defense against microbial invasion, and also regulates cell proliferation, immune activity and angiogenesis. Previous studies have reported that LL-37 participates in the development of numerous tumour types, such as ovarian cancer, lung cancer, melanoma and breast cancer. However, the function of LL-37 in the development of skin squamous cell carcinoma (SCC) has not yet been fully elucidated. The aim of the current study was to investigate how LL-37 promotes the expression of Y-box binding protein 1 (YB-1) in SCC. Short interfering RNA (siRNA) was used to inhibit the expression of YB-1, and
Sigma-Aldrich
MISSION® esiRNA, targeting human YBX1
Pricing and availability is not currently available.