MilliporeSigma
HPLC Analysis of Anthocyanins in Red Wine on Ascentis® C18
HPLC Analysis of Anthocyanins in Red Wine on Ascentis® C18 application for HPLC
Materials
Related product
Product No.
Description
Pricing
Ascentis® C18 HPLC Column
5 μm particle size, L × I.D. 25 cm × 4.6 mm
CONDITIONS
column
Ascentis C18, 25 cm × 4.6 mm I.D., 5 μm particles (581325-U)
mobile phase
[A] water:formic acid (9:1), [B] acetonitrile:water:formic acid (5:4:1)
flow rate
1 mL/min
column temp.
25 °C
detector
VIS, 518 nm
injection
10 μL
sample
red wine diluted with water, centrifuged and passed through a 0.45 μm filter
Description
Legal Information
Ascentis is a registered trademark of Merck KGaA, Darmstadt, Germany