Skip to Content
MilliporeSigma

396451

4-[4-(Trifluoromethyl)phenoxy]phenol

98%

Sign In to View Organizational & Contract Pricing.

About This Item

Linear Formula:
CF3C6H4OC6H4OH
CAS Number:
Molecular Weight:
254.20
UNSPSC Code:
12352100
NACRES:
NA.22
PubChem Substance ID:
EC Number:
254-549-8
Beilstein/REAXYS Number:
2055876
MDL number:

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Product Name

4-[4-(Trifluoromethyl)phenoxy]phenol, 98%

InChI key

FJFSSESWAFPCSU-UHFFFAOYSA-N

InChI

1S/C13H9F3O2/c14-13(15,16)9-1-5-11(6-2-9)18-12-7-3-10(17)4-8-12/h1-8,17H

SMILES string

Oc1ccc(Oc2ccc(cc2)C(F)(F)F)cc1

assay

98%

form

solid

mp

49-54 °C (lit.)

functional group

fluoro

Looking for similar products? Visit Product Comparison Guide

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Erik R Christensen et al.
Environmental toxicology and chemistry, 28(4), 826-835 (2009-04-28)
We derive equations for the effective concentration giving 10% inhibition (EC10) with 95% confidence limits for probit (log-normal), Weibull, and logistic dose-response models on the basis of experimentally derived median effective concentrations (EC50s) and the curve slope at the central
Tukayi Kudanga et al.
Bioresource technology, 101(8), 2793-2799 (2010-01-02)
This work presents for the first time the mechanistic evidence of a laccase-catalyzed method of covalently grafting hydrophobicity enhancing fluorophenols onto Fagus sylvatica veneers. Coupling of fluorophenols onto complex lignin model compounds guaiacylglycerol beta-guaiacyl ether and syringylglycerol beta-guaiacyl ether was

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service