Skip to Content
MilliporeSigma

401501

Germanium(IV) fluoride

99.95%

Synonym(s):

Germanium tetrafluoride

Sign In to View Organizational & Contract Pricing

About This Item

Linear Formula:
GeF4
CAS Number:
Molecular Weight:
148.63
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.23

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

assay

99.95%

mp

−36.5 °C (lit.)

transition temp

sublimation point -36.5 °C

density

2.46 g/mL at -37 °C (lit.)

SMILES string

F[Ge](F)(F)F

InChI

1S/F4Ge/c1-5(2,3)4

InChI key

PPMWWXLUCOODDK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Germanium(IV) fluoride in combination with disilane gas has recently been used to fabricate SiGe crystallites directly on glass substrates. Intense visible light emission from Ge and Si nanocrystals is of particular interest because of potential uses in optoelectronic devices.

Packaging

Packaged in carbon steel lecture bottles with CGA 110F/180M 316ss diaphragm valve
Packaged in electropolished stainless steel lecture bottle with a stainless steel valve and CGA 330 outlet.

Other Notes

Monel control valve Z146978 or Monel gas regulator Z562483 is recommended.
Requires 316 SS gas regulator Z173959 or Z173967.

pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Skin Corr. 1B

Storage Class

2A - Gases

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service