Skip to Content
MilliporeSigma

APO414278590

Apollo Scientific Ltd

3-Chloro-4-fluorobenzhydrazide

Sign Into View Organizational & Contract Pricing

Select a Size

Product APO414278590 is currently not available for purchase. Contact Technical Service

About This Item

CAS Number:
Molecular Weight:
188.59
MDL number:

Skip To

Iupac Name

3-chloro-4-fluorobenzohydrazide

Inchi Code

1S/C7H6ClFN2O/c8-5-3-4(7(12)11-10)1-2-6(5)9/h1-3H,10H2,(H,11,12)

InChI key

UAXDFZDWRYDZEJ-UHFFFAOYSA-N

storage temp.

Ambient Temperature

Country of Origin

GB

Shipping Temperature

Ambient Temperature

Physical Form

Solid

Product Link

https://store.apolloscientific.co.uk/product/3-chloro-4-fluorobenzhydrazide

Synonym

3-Chloro-4-fluorobenzohydrazide

pictograms

Exclamation mark

Signal Word

Warning

Precautionary statements

MSDS

https://store.apolloscientific.co.uk/product/3-chloro-4-fluorobenzhydrazide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library