Sign Into View Organizational & Contract Pricing
Select a Size
Product LIF388583915 is currently not available for purchase. Contact Technical Service
About This Item
Skip To
Iupac Name
6-oxo-3,4-dihydro-2H,6H-pyrimido[2,1-b][1,3]thiazine-3-carboxylic acid hydrochloride
Inchi Code
1S/C8H8N2O3S.ClH/c11-6-1-2-9-8-10(6)3-5(4-14-8)7(12)13;/h1-2,5H,3-4H2,(H,12,13);1H
InChI key
PTWRMZKWXRDMTJ-UHFFFAOYSA-N
Purity
95%
Country of Origin
UA
Physical Form
solid
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.