About This Item
Skip To
Product Name
Anti-BAF180 Antibody, clone 370.1.8C3, clone 370.1.8C3, from rat
biological source
rat
conjugate
unconjugated
antibody form
purified immunoglobulin
antibody product type
primary antibodies
clone
370.1.8C3, monoclonal
species reactivity
human
packaging
antibody small pack of 25 μL
technique(s)
western blot: suitable
isotype
IgG2aκ
NCBI accession no.
UniProt accession no.
target post-translational modification
unmodified
Gene Information
human ... PBRM1(55193)
1 of 4
This Item | ABE70 | AMAB90690 | HPA015629 |
|---|---|---|---|
| biological source rat | biological source rabbit | biological source mouse | biological source rabbit |
| clone 370.1.8C3, monoclonal | clone polyclonal | clone CL0331, monoclonal | clone polyclonal |
| Gene Information human ... PBRM1(55193) | Gene Information human ... PBRM1(55193) | Gene Information human ... PBRM1(55193) | Gene Information human ... PBRM1(55193) |
| antibody form purified immunoglobulin | antibody form purified antibody | antibody form purified immunoglobulin | antibody form affinity isolated antibody |
| species reactivity human | species reactivity human, mouse, rat | species reactivity human | species reactivity mouse, rat, human |
| UniProt accession no. | UniProt accession no. | UniProt accession no. | UniProt accession no. |
Analysis Note
Western Blotting Analysis: A 1:1,000 dilution of this antibody detected BAF180 in HEK293 cell lysate.
Application
Epigenetics & Nuclear Function
Biochem/physiol Actions
Disclaimer
General description
Immunogen
Other Notes
Physical form
Preparation Note
Not finding the right product?
Try our Product Selector Tool.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service


