About This Item
Skip To
Product Name
Anti-CD45.2 Antibody (mouse), PE-Cy7, clone 104, clone 104, 0.2 mg/mL, from mouse
biological source
mouse
conjugate
PE-Cy7
antibody form
purified antibody
antibody product type
primary antibodies
clone
104, monoclonal
species reactivity
mouse
concentration
0.2 mg/mL
technique(s)
flow cytometry: suitable
immunocytochemistry: suitable
immunoprecipitation (IP): suitable
isotype
IgG2aκ
UniProt accession no.
shipped in
wet ice
target post-translational modification
unmodified
Quality Level
Gene Information
mouse ... Ptprc(19264)
Related Categories
1 of 4
This Item | MABF438 | MABF435 | MABF596 |
|---|---|---|---|
| biological source mouse | biological source mouse | biological source mouse | biological source mouse |
| conjugate PE-Cy7 | conjugate PE | conjugate Allophycocyanin conjugate | conjugate FITC conjugate |
| species reactivity mouse | species reactivity mouse | species reactivity mouse | species reactivity mouse |
| antibody form purified antibody | antibody form purified antibody | antibody form purified antibody | antibody form purified antibody |
| Gene Information mouse ... Ptprc(19264) | Gene Information mouse ... Ptprc(19264) | Gene Information mouse ... Ptprc(19264) | Gene Information mouse ... Ptprc(19264) |
| clone 104, monoclonal | clone 104, monoclonal | clone 104, monoclonal | clone 104, monoclonal |
Analysis Note
Flow Cytometry Analysis: 0.25 µg of this antibody detected CD45.2 in one million C57Bl/6 splenocytes.
General description
Not finding the right product?
Try our Product Selector Tool.
Storage Class
12 - Non Combustible Liquids
wgk
nwg
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service


