Skip to Content
MilliporeSigma

76850

1,2,2,6,6-Pentamethylpiperidine

purum, ≥99.0% (GC)

Synonym(s):

PMP, Pempidine

Sign In to View Organizational & Contract Pricing

Select a Size

Pricing and availability is not currently available.

About This Item

Empirical Formula (Hill Notation):
C10H21N
CAS Number:
Molecular Weight:
155.28
Beilstein/REAXYS Number:
103806
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

grade

purum

assay

≥99.0% (GC)

refractive index

n20/D 1.460 (lit.)
n20/D 1.460

bp

187-188 °C (lit.)

density

0.858 g/mL at 25 °C (lit.)

SMILES string

CN1C(C)(C)CCCC1(C)C

InChI

1S/C10H21N/c1-9(2)7-6-8-10(3,4)11(9)5/h6-8H2,1-5H3

InChI key

XULIXFLCVXWHRF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

One of the most strongly basic tertiary amines (sterically hindered and nonquaternizing)[1]

pictograms

FlameSkull and crossbones

signalword

Danger

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

131.0 °F - closed cup

flash_point_c

55 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H.Z. Sommer et al.
The Journal of Organic Chemistry, 36, 824-824 (1971)
M L Kemel et al.
Neuroscience, 50(3), 597-610 (1992-10-01)
Previously, using a new in vitro microsuperfusion procedure, we have demonstrated marked differences in the cholinergic presynaptic regulation of the release of [3H]dopamine continuously synthesized from [3H]tyrosine in two close striosomal- and matrix-enriched areas of the cat caudate nucleus. A
[Tuberculous osteoarthritis revealed by ankle injury].
O Louletzian et al.
Presse medicale (Paris, France : 1983), 28(10), 531-532 (1999-04-21)
Dean A Myers et al.
Behavioural brain research, 161(1), 39-44 (2005-05-21)
Corticosterone (CORT) micropellets were stereotaxically placed bilaterally at the dorsal margin of the central nucleus of the amygdala (CeA). Both behavioral and physiological responses were recorded (plus maze and colonic discomfort) at 7 days post-implantation. Corticosterone reduced the exploration of
V E Gmiro et al.
Eksperimental'naia i klinicheskaia farmakologiia, 63(2), 16-20 (2000-06-02)
Pyrilene acts as a central H-cholinoblocker upon intramuscular injection at a dose of 0.02-0.08 mg/kg, and as an NMDA-blocker, when the dose is increased to 0.2-0.8 mg/kg. Similarly, amantadine exhibits the properties of H-cholinoblocker and NMDA-blocker in the dose intervals

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service