Sign In to View Organizational & Contract Pricing.
About This Item
Empirical Formula (Hill Notation):
C29H48N12O8
CAS Number:
Molecular Weight:
692.77
UNSPSC Code:
12352200
Skip To
assay
≥98% (HPLC)
form
lyophilized powder
storage condition
desiccated
protect from light
color
white
solubility
H2O: soluble >1 mg/mL
storage temp.
−20°C
Amino Acid Sequence
Gln-Arg-Phe-Ser-Arg
Biochem/physiol Actions
Opiorphin is a natural analgesic, dual action pain killer; six times stronger than the opiate morphine.
Opiorphin is a natural analgesic, secreted into human saliva. Opiorphin demonstrates dual-inhibitory potency on the enkephalin-inactivating ectopeptidases human NEP (hNEP) and human AP-N (hAP-N). As an analgesic, Opiorphin is six times stronger than the opiate morphine. Opiorphin is closely related to the rat sialorphin peptide, which is an inhibitor of pain perception and acts by potentiating endogenous micro- and δ-opioid receptor-dependent enkephalinergic pathways. In rat studies, Opiorphin suppressed pain sensation for both chemical-induced inflammation and acute physical pain. In both cases, the administered dose of 1 mg/kg provided the same painkilling power as 3-6 mg/kg of morphine.
Features and Benefits
The peptide content is 78.9%
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service