Skip to Content
MilliporeSigma

TA9H98048AD9

Targetmol Chemicals Inc.

3-(2-aminopropyl)phenol hydrochloride

Synonym(s):

3-(2-aminopropyl)phenol hydrochloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Product TA9H98048AD9 is currently not available for purchase. Contact Technical Service

Skip To

Purity

99.64%

Inchi Code

1S/C9H13NO.ClH/c1-7(10)5-8-3-2-4-9(11)6-8;/h2-4,6-7,11H,5,10H2,1H3;1H

Inchi Key

PVIAWJZKJCBTTA-UHFFFAOYSA-N

Storage temp.

-80/-20

Country of Origin

CN

Shipping Temperature

4

Physical Form

solid

MSDS

https://www.targetmol.com/compound/3-(2-aminopropyl)phenol hydrochloride


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library