MilliporeSigma
  • Transfected plasmid DNA is incorporated into the nucleus via nuclear envelope reformation at telophase.
Transfected plasmid DNA is incorporated into the nucleus via nuclear envelope reformation at telophase.
Communications biology (2022-01-22)
Tokuko Haraguchi, Takako Koujin, Tomoko Shindo, Şükriye Bilir, Hiroko Osakada, Kohei Nishimura, Yasuhiro Hirano, Haruhiko Asakawa, Chie Mori, Shouhei Kobayashi, Yasushi Okada, Yuji Chikashige, Tatsuo Fukagawa, Shinsuke Shibata, Yasushi Hiraoka
ABSTRACT
DNA transfection is an important technology in life sciences, wherein nuclear entry of DNA is necessary to express exogenous DNA. Non-viral vectors and their transfection reagents are useful as safe transfection tools. However, they have no effect on the transfection of non-proliferating cells, the reason for which is not well understood. This study elucidates the mechanism through which transfected DNA enters the nucleus for gene expression. To monitor the behavior of transfected DNA, we introduce plasmid bearing lacO repeats and RFP-coding sequences into cells expressing GFP-LacI and observe plasmid behavior and RFP expression in living cells. RFP expression appears only after mitosis. Electron microscopy reveals that plasmids are wrapped with nuclear envelope (NE)‒like membranes or associated with chromosomes at telophase. The depletion of BAF, which is involved in NE reformation, delays plasmid RFP expression. These results suggest that transfected DNA is incorporated into the nucleus during NE reformation at telophase.
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, clone DM1A, ascites fluid
Pricing and availability is not currently available.
Sigma-Aldrich
Anti-LEMD2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Pricing and availability is not currently available.
Sigma-Aldrich
Nocodazole, ≥99% (TLC), powder
Pricing and availability is not currently available.