MilliporeSigma
Germline
NPJ genomic medicine (2017-12-22)
Lamis Yehia, Ying Ni, Charis Eng
ABSTRACT
Bannayan-Riley-Ruvalcaba syndrome (BRRS) is a rare congenital disorder classically characterized by macrocephaly in combination with intestinal hamartomatous polyposis, vascular malformations, lipomas, and genital lentiginosis. Germline
Sigma-Aldrich
Thallium(III) nitrate trihydrate
Pricing and availability is not currently available.