Test Strips, Papers & Readers
Test strips are a simple, effective, and low-cost analytical tool used for the rapid detection of an analyte in the field or on-site. A test strip coated with a chemical that reacts with the analyte is either dipped in the sample solution or the sample is loaded onto the test strip. The use of test strips for in-process detection is advantageous due to the ease of use and the high specificity offered by them. The results of test strips are either based on visual observation or the quantitative analysis using digital readers.
Related Resources
- Catalog: Water, Environmental, and Food & Beverage Analysis
We created an easy-to-navigate catalog to help you quickly find everything required for your measurements.
- Brochure: RQflex® 20 Reflectometer - For Rapid and Precise In-process Monitoring and Rapid QC Analysis
The Reflectoquant® system consists of test strips and reflectometers. This mobile laboratory gives you all the tools you need for high quality analyses for a wide spectrum of sample materials from food, water, and industrial to environmental samples.
- Brochure: Soil Testing for Cannabis Growers: Applications
As a leading supplier to the global Life Science industry, we understand the analytical and regulatory challenges involved in cultivation, processing, extraction, and quality control of the cannabis products. We offer expertise and a range of products and services to support your cannabis testing workflow.
- Application Note: Reflectometry – Hydroxymethylfurfural (HMF) in Honey
Hydroxymethylfurfural (HMF) in honey – reflectometric determination after after conversion with a barbituric acid derivative and an aminophenazone derivative.
- Application Note: Nitrate in Soil with MQuant® StripScan App
The following application note describes the determination of the nitrate concentrations in soil samples using the MQuant® Nitrate test strips in combination with the MQuant® StripScan App.
- MQuant® StripScan Webportal
- White Paper: Beverage Testing with the MQuant® StripScan Mobile App
This white paper examines the use and efficiency of the MQuant® StripScan mobile app to measure the pH of orange juice samples and nitrate concentration of mineral water samples.
- Nitrate Detection in Wastewater
Reflectometric determination of nitrate in wastewater using Griess reagent and Reflectoquant® test strips.
- pH in Coffee
Explore the cost-effective and reliable smartphone-based pH determination of coffee samples using MQuant® pH test strips and the MQuant® StripScan App.
- pH of White Wine
Rapid and simple smartphone-based determination of the pH value of wine (white) after reaction with chemically bound pH indicator strips.
- pH in Fruit Juices
An easy-to-use, economical, and smartphone-based reliable determination of pH in fruit juices using the MQuant® pH test strips and the MQuant® StripScan app.
- pH in Soft Drinks
An easy-to-use and economical approach for reliable determination of pH of soft drinks with MQuant® pH test strips and MQuant® StripScan App.
Related Webinars
Get accurate test results with your handheld instrument or smartphone.
Disinfection control within production facilities, particularly within the food and beverage industry, is critical to ensure product safety and meet regulatory compliance.
Test Strips in Food & Beverage Applications
Our selection of test strips for food and beverage applications includes MQuant® and Reflectoquant® test strips for visual or instrumental readout. These can be easily used anywhere and can significantly reduce the time and cost of the laboratory analysis. MQuant® test strips are used for the semiquantitative screening of glucose, lactoperoxidase in pasteurized milk, and detection of ascorbic acid and lactic acid in fruits, among others. The Reflectoquant® test strips are used in combination with the RQflex® 20 reflectometer for the easy and accurate quantitative measurement of different substances in food and beverage production as well as water testing. Reflectoquant® test strips are available to determine for instance the hydroxymethylfurfural (HMF) content in honey and fruit juices, total sugar content in potatoes, and nitrate content in vegetables.
MQuant® Test Strips for Semiquantitative Testing
MQuant® test strips offer semiquantitative detection of more than 30 different ions, organic, and inorganic compounds in the mg/L to g/L concentration range. MQuant® test strips feature minute amounts of the reagent on an absorbent pad mounted to a polyethylene terephthalate (PET) film. Next to the applications in food & beverage mentioned above, MQuant® test strips can be used for different semiquantitative measurements, including:
- Potassium content of drinking water, mineral water, industrial water, wastewater, soil samples, wine, beer, and fruit juices
- Arsenic in water, soil extracts, pharmaceutical products, prepared biological specimens, and liquid foods
- Nitrite in boiler and cooling water
- Tin and tin compounds in electroplating baths, reducing agents, stabilizers, catalysts, disinfectants, fungicides, and mordants used in the textile industry
Reflectoquant® Test Strips and Reader for Quantitative Testing
Our Reflectoquant® series includes Reflectoquant® test strips and the RQflex® 20 instrument for on-site, direct sample analysis in food and beverage production and water testing. Reflectoquant® test strips are barcoded with batch-specific calibration to offer reliable, quantitative results in minutes. Next to the applications in food & beverage mentioned above, Reflectoquant® test strips can be used for many different quantitative measurements including:
- Malic acid in wines
- Formaldehyde in fish products
- Calcium in cheese
- Peroxide and Peracetic acid in disinfection control
MQuant® StripScan App for Digital Readout of MQuant® Test Strips
Our MQuant® StripScan app allows the easy readout of nitrate and pH results in seconds. The app provides effortless result analysis, management, and data sharing along with efficient visualization for easy and transparent data interpretation.