This product is prepared as a 0.0025% TrypZean solution. A prepared solution at a higher concentration is not currently available. Please refer to the powder form, product T3568, which has a minimum activity specification of 3350 units per milligram. The product is tested for solubility at 50,000 u/mL in water and may be used to prepare solutions at higher concentrations. Prepared stock solutions require filter-sterilization prior to use. Please see the link below to review this product:
https://www.sigmaaldrich.com/product/sigma/t3568
Select a Size
| Size/SKU | Availability | Price |
|---|---|---|
100 mL | Check Cart for Availability | $51.90 |
500 mL | Check Cart for Availability | $207.00 |
About This Item
recombinant
expressed in corn
Quality Segment
sterility
sterile-filtered
form
aqueous solution
concentration
1 ×
technique(s)
cell culture | mammalian: suitable
pH
7.70-8.30
shipped in
dry ice
storage temp.
−20°C
General description
Application
Features and Benefits
- Animal component-free – eliminates the risk of viruses, BSE, or other potential adventitious agents
- TrypZean is recombinant trypsin – using the same enzyme (with the same kinetics) for cell detachment means minimal protocol changes
- Enzyme inhibition – soybean trypsin inhibitor and other inhibitors work the same with TrypZean as they do with native trypsins (weight-to-weight basis)
- High purity – TrypZean provides increased specificity and eliminates contaminating activities found in lower purity enzymes
- Convenience – TrypZean solution is a sterile-filtered solution formulated at the optimal concentration to dissociate adherent cell lines
Legal Information
1 of 1
This Item | |||
|---|---|---|---|
| description recombinant, expressed in corn, sterile-filtered | description recombinant, expressed in corn, lyophilized powder, ≥3,350 units/mg solid | description sterile-filtered, BioReagent, suitable for cell culture, 25 g porcine trypsin per liter in Hanks′ Balanced Salt Solution with phenol red | description sterile-filtered, BioReagent, suitable for cell culture, 25 g porcine trypsin per liter in 0.9% sodium chloride |
| technique(s) cell culture | mammalian: suitable | technique(s) - | technique(s) cell culture | mammalian: suitable | technique(s) cell culture | mammalian: suitable |
| form aqueous solution | form lyophilized powder | form solution | form solution |
| concentration 1 × | concentration - | concentration - | concentration - |
| recombinant expressed in corn | recombinant expressed in corn | recombinant - | recombinant - |
| shipped in dry ice | shipped in - | shipped in dry ice | shipped in dry ice |
| sterility sterile-filtered | sterility - | sterility sterile-filtered | sterility - |
Storage Class
12 - Non Combustible Liquids
WGK
nwg
Flash Point (°F)
Not applicable
Flash Point (°C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
-
Bonjour, existe t-il une autre concentration pour la trypzean ?
1 answer-
Helpful?
-
-
What is the working concentration of recombinant trypsin used ?
1 answer-
Product T3449 is prepared from a 0.0025% concentration.
Helpful?
-