User Guide Millivac™ Mini Vacuum Pump
Introduction
The Millivac™ Mini Vacuum Pump is a compact, lightweight, and easy to use pump, designed for small volume/aqueous flask filtration applications. This diaphragm pump has premium, corrosion-resistant wetted parts and is fitted with a patented multi-port valve system that increases performance and tolerates liquids accidentally drawn into the system without loss of performance. The fan-cooled, ball-bearing motor is permanently lubricated and has a single-use (fuse) overload protector. The Millivac™ Mini Vacuum Pump is supplied with tubing and Millex®-FG50 filters
CAUTION: Always use the supplied Millex®-FG50 filter or a vacuum-flask water trap in conjunction with the pump, as indicated in Figures 1 and 2. Use this pump to pump air or gas only, not liquids or particulates.
Rules for Safe Operation
  • Do NOT pump flammable or explosive gases or vapors or operate this pump in or near an area containing flammable or explosive gases or vapors.
  • Do NOT operate in an area where aerosol (spray) products are being used.
  • Do NOT operate this product near flames.
  • For indoor use only.
  • Do not operate in an area where the pump can fall or be pulled into water or any other liquids.  Do not reach for this product if it has fallen into liquid. Unplug immediately.
CAUTION
  • Inspect hose, plug, and cord for signs of damage before use. Do not use if a deficiency is found. Never operate a unit that has been dropped, damaged or has fallen into water. Contact us for information; see Technical Assistance section for details.
  • Keep cord away from heated surfaces.
  • Do not block the ventilation ports located on the housing and keep them free of dirt and foreign objects.
  • The pump has a single-use (fuse) type of overload protector. If the motor overheats, the protector will shut the unit off. If this occurs, unplug the pump and contact Technical Service.
  • Use only to pump air or gas, not liquids or particulates. Damage to the pump or loss of performance can occur if large amounts of liquid or particulates enter the system. The life of the pump can be prolonged if the formation of condensate within the pump is avoided. The vacuum trap should be emptied after each use.
Installation and Usage Guidelines
  1. The vacuum pump should be operated in a clean, dry, well-ventilated area, in a location where the temperature remains between 5–40 °C (41–104 °F). This is particularly important when the unit is installed in a confined space. Protect external parts from direct or indirect moisture contact. Avoid operating the pump in very dusty conditions.
  2. To prevent liquids or vapors from entering the pump when applying vacuum, install the Millex®-FG50 filter in-line between the filter flask and pump (Figure 1). Alternatively, the pump can be protected by a flask vacuum trap consisting of a 1 liter filtering flask (cat. no. XX1014705), No. 8 stopper (cat. no. XX2014718) and silicone tubing (Figure 2). When using the flask vacuum trap, it is not necessary to use the Millex® filter. For best results, keep the tubing between the pump and equipment as short as possible. Additional tubing is available as cat. no. XX7100004.
Figure 1. Millex®-FG50 filter pump protection
Figure 2. Vacuum trap pump protection
  1. Replace the Millex®-FG50 filter (cat. no. SLFG05010) if it gets wet or appears to be plugged.
  2. When the unit is not in use, wrap the power cord around the vacuum/pressure pump and store in a dry place. Do not abuse the cord.
  3. This pump is 100% oil-free. All bearings are sealed and permanently lubricated. Lubrication should not be attempted.
  4. Be sure that the available electric power matches specifications of the electric motor listed on the identification plate. Serious damage may occur to the motor if connected to an improper voltage. This pump must be grounded. In the event of an electrical short circuit, grounding reduces the risk of electric shock. Do not remove the ground pin from the plug.
  5. This product is equipped with a cord having a grounding wire with an appropriate grounding plug. The plug must be plugged into an outlet that is properly installed and grounded in accordance with all local electrical codes and ordinances.
  6. This pump is designed to start against atmospheric pressure only, not under any vacuum load. Care must be taken to eliminate load after pump is turned off.
  7. Output flow should not be throttled or restricted for any reason.
  8. Be sure that the pump is installed at the highest point within the system to prevent large quantities of condensate from entering the unit.
  9. To avoid damaging polyphenylene sulfide (PPS) heads, use only plastic hose barb fittings.
How to Use the Millivac™ Mini Vacuum Pump
The Millivac Mini Vacuum Pump is shipped ready for use. See Installation and Usage and Specifications sections for required operating conditions.
  1. Remove any protective plastic plugs supplied in the intake or pressure ports of the pump prior to applying power to the motor.
  2. Plug pump into appropriate electrical outlet. Turn pump on with On/Off switch on front of unit.
  3. Run the pump for a few minutes to warm it up before handling saturated or nearly saturated vapors.
  4. Attach system tubing to the hose barb on the right of the pump top for vacuum applications.
  5. After use, let the pump run for about 2 minutes with room air before switching it off, to purge out droplets of liquid that may have formed on the inside of the pump. This prevents crystallization and/or absorption of liquids by the pump materials, and prolongs pump life.
Troubleshooting
Maximum vacuum is not reached
  1. Confirm that all system connections are gas-tight. To detect leaks, slightly pressurize the system and paint the suspected area with a thick soap solution. Escaping air will produce soap bubbles.
  2. Be sure that all screws on the head assembly are snug. To avoid damaging pump, do not overtighten screws.
  3. Disassemble the valve assembly as described in “Maintenance”. Look for any foreign matter, commonly bits of polytetrafluoro-ethylene (PTFE) tape or particulates carried into the valve system or crystallized residue from previously pumped vapors. All of the above must be cleaned out and the pump reassembled with clean or new parts.
Pump does not run and pump On/Off switch is dark
  1. Confirm that power is being supplied to the pump from the power source and the pump switch is in the On position.
  2. An overcurrent or other problem may have blown the fuses. The fuse holder is located next to power socket. Before checking fuses:
    WARNING: Shut off all power to pump and unplug pump's power cord from electrical source.
  3. Pull out the fuse holder while depressing the small tab, and determine if the fuses are good. The proper replacement fuses should always be used. Consult the nameplate or call Technical Service for the proper rating and type of fuse.
Pump does not run and pump On/Off switch is lit
WARNING: Motor is thermally protected and will automatically restart unexpectedly when overload device resets.
  1. It is possible that the thermal protective device has activated. Turn off the pump and let it cool for 15 minutes before trying again.
  2. If the pump starts, determine the cause of the overload before using the pump. Possible causes are:
    • Electrical power source does not match motor requirements
    • Insufficient ventilation of the motor caused by blockage or dirt
    • Defective pump assembly or motor
    • Application is exceeding the pressure rating of the pump
Maintenance
Under normal operating conditions, and using proper handling procedures, the Millivac™ Mini Vacuum Pump will provide hours of service-free operation. It requires no lubrication and the heads and valves can be easily disassembled for occasional cleaning and/or replacement of valve plate, seal ring, and diaphragm. Leakage and contamination are the general causes of problems associated with poor vacuum pressure. To operate at maximum efficiency, the pump must be thoroughly clean. If the system is completely clean and free from leaks, and unwarranted vacuum problems still exist, the valve plate, seal ring, and diaphragm may need to be replaced. Refer to instructions below for cleaning and part replacement.
Cleaning and Replacement of Valve Plate, Seal Ring, and Diaphragm (refer to Expanded View of Pump Head)
Materials needed: Replacement kit XF5423055 (includes 1 structured diaphragm, 2 valve plates, and 2 seal rings)
Tools required: Small Phillips-head screwdriver, marking pencil, mild solvent to clean residue from heads
Warning: Shut off all power to pump before disassembly. Unplug pump's power cord from electrical source.
  1. Be sure pump is dry and free of hazardous materials by operating it for 5 minutes in free air.
  2. Mark the relative position of the head parts with a pencil line to ensure correct assembly later. It is not necessary to remove the hose barbs.
  3. Remove the 4 perimeter head screws B on the pump head.
  4. Lift the head assembly and rubber skirt straight up and place on a clean, flat work surface.
  5. Carefully separate the headplate A from the intermediate plate C. Two seal rings and two valve plates are now exposed.
  6. Remove seal rings E and valve plate D. Carefully clean any residue from the valve seats with a mild solvent. Do not scratch the PPS surfaces. The area around the valve plate must be smooth and dry. If pitting of the pump parts or tearing of the diaphragm is observed, it is possible that the gas/vapor being pumped is attacking the wetted parts of the pump. Chemical resistance charts should be consulted if you are in doubt. Generally, replacement of the diaphragm, seal rings and valve plates will restore the pump to operating specifications.
  7. Install new seal rings E and valve plates D from the spare parts kit, if required. Skip to step 11 to reassemble head without replacing diaphragm.
To replace the diaphragm:
NOTE: There are a number of small shim rings J located between the diaphragm and the diaphragm support cup G and between the diaphragm support cup G and the connecting rod M. Use care when disassembling the diaphragm so that you can note the quantity, position, and thickness of the shim rings. These shim rings must be reinstalled in the original quantity and position for proper pump operation (see expanded view).
  1. Carefully lift the diaphragm F edge from the circular groove, and begin to unscrew the diaphragm from connecting rod M by hand. Before completely unscrewing the diaphragm, flip the pump upside down and complete removal of the diaphragm. This will prevent the small shims on the diaphragm’s threaded stud from falling into the chamber.
  2. Hint: It may be easier to perform the following step by holding the pump upside down, or placing it on its switch end on a flat surface before screwing the assembled parts into connecting rod M. Assemble all of the components according to the diagram, over the threaded stud end of replacement diaphragm F and align the stud with the threaded hole of connecting rod M in the chamber. Be sure all parts are centered. Screw the new diaphragm into the connecting rod by hand. Do not overtighten.
  3. Once installed, press down on the diaphragm until the lip on the edge of the diaphragm fits into the groove on the rim of the mounting flange.
  4. Reassemble the headplate A (note guide pin), and intermediate plate C in accordance with the mark you made in step 2.
  5. Replace the pump head assembly and tighten head screws B snugly in a crosswise pattern. Do not overtighten screws.
  6. Replace rubber protective boot.
Specifications
Performance
Vacuum Pressure 10 kPa absolute (3.0 in. Hg absolute, 27 in. Hg gauge)
Dimensions
Length 22.6 cm (8.9 in.)
Width 9 cm (3.5 in.)
Height 14.1 cm (5.6 in.)
Weight (approximate) 1.9 kg (4.2 lb)
Tubing Connectors 6 mm (1/8 in. NPT) x hose barb for 6.4 mm (1/4 in.) ID tubing
Materials of Construction
Pump Wetted Parts Polyphenylene sulfide (PPS) head, ethylene propylene diene monomer (EPDM) diaphragm, fluorinated polypropylene monomer (FPM) valves and seals
Tubing Connectors Polyethylene
Electrical Specifications
115 V (60 Hz) 3 conductor (grounded) power cord; protection class IP20