MilliporeSigma
HomeSingle Molecule Counting (SMC®) TechnologyOptimize Sample Performance with the SMC® Plate Washer Evaluation Kit
Optimize Sample Performance with the SMC® Plate Washer Evaluation Kit
With SMC® technology, immunoassays reach sensitivities in the femtogram/mL level, meaning they require both the highest performance and the highest consistency through each step of your assay workflow. Before running your precious samples, learn how to evaluate the performance of your plate washer using the SMC® Plate Washer Evaluation Kit.
What is the SMC® Plate Washer Evaluation Kit?
The SMC® Plate Washer Evaluation Kit provides all the reagents required to run 5 plates, meaning that you can test multiple washers or test before and after plate washer maintenance. It also includes a unique reagent that mimics steps in the SMC® immunoassay, with an abbreviated workflow that both isolates and highlights the performance of the plate washer. This reagent was optimized to analyze the BioTek® 405™TSUVS Plate Washer for SMC® and MILLIPLEX® Assays and to analyze data on the SMCxPRO® immunoassay platform. With this kit, you can confirm optimal plate washer performance before evaluating your precious samples.
Plate Washer Evaluation Kit Workflow
As mentioned, this plate washer evaluation kit has an abbreviated workflow to execute assays in approximately 1 hour. The steps in the workflow help to minimize sources of error in assay execution and determine intra-plate precision.
This workflow is shown below and has 8 steps.
The steps in THE SMC® PLATE WASHER EVALUATION KIT workflow include:
Add 200 µL/well of pre-formed complex to 96-well plate
Add 200 µL/well of pre-formed complex to 96-well plate
    Plate washer protocols
    a.    Post capture wash (~8 minutes)
    b.    4 cycle pre-transfer (~10 minutes)
    c.     Final aspirate (~2 minutes)
    d.    Remove the plate from the washer
      Add 10 µL of elution buffer/well
      Then, incubate, shaking for 10 minutes
        Place the plate on the magnet for 2 minutes
          Transfer of 10 µL eluate to SMC<sup>®</sup> 384-well reading plate
          The plate should contain 10 µL of Buffer D
            Place the plate in the SMCxPRO® instrument to analyze
              Fluors are excited
                Obtain data
                  Learn more about tips for the SMCxPRO® platform in our SMCxPRO® Best Practices article.
                  Data from a Plate Washer Before and After Preventive Maintenance
                  The first data set is from a plate washer in need of preventative maintenance (Table 1). The highlighted wells, with lower Response Event values, indicate microparticle loss that will lead to poor assay precision. Nearly all columns and rows have greater than 20% CV.
                  1
                  2
                  3
                  4
                  5
                  6
                  7
                  8
                  9
                  10
                  11
                  12
                  Row
                  Mean
                  SD
                  CV
                  A
                  57
                  41
                  69
                  41
                  84
                  63
                  55
                  92
                  90
                  87
                  73
                  100
                  71
                  20
                  28
                  B
                  87
                  16
                  26
                  2
                  36
                  43
                  32
                  34
                  30
                  42
                  41
                  89
                  40
                  25
                  64
                  C
                  88
                  38
                  18
                  24
                  40
                  31
                  15
                  36
                  71
                  69
                  81
                  91
                  50
                  28
                  56
                  D
                  82
                  60
                  25
                  11
                  29
                  46
                  54
                  44
                  51
                  52
                  62
                  79
                  50
                  21
                  42
                  E
                  55
                  6
                  46
                  13
                  33
                  34
                  38
                  7
                  40
                  47
                  53
                  70
                  37
                  20
                  54
                  F
                  59
                  14
                  49
                  34
                  25
                  18
                  1
                  2
                  61
                  55
                  61
                  72
                  38
                  25
                  67
                  G
                  42
                  10
                  11
                  1
                  25
                  8
                  3
                  32
                  8
                  29
                  54
                  69
                  24
                  22
                  89
                  H
                  55
                  18
                  69
                  23
                  68
                  44
                  63
                  69
                  78
                  63
                  84
                  75
                  59
                  21
                  35
                  Column Mean
                  66
                  25
                  39
                  19
                  43
                  36
                  33
                  39
                  54
                  56
                  64
                  81
                  SD
                  18
                  19
                  23
                  14
                  22
                  17
                  24
                  30
                  27
                  18
                  15
                  11
                  CV
                  27
                  75
                  58
                  78
                  51
                  48
                  74
                  76
                  50
                  32
                  23
                  14
                  Table 1. Plate washer before preventative maintenance.
                  Wells in orange indicate wells with visible loss of microparticles. CVs in pink are >20% CV.
                  The second data set is the same plate washer assessed after performing recommended maintenance procedures (Table 2).
                  The overall plate, column, and row precision have decreased to below 20% CV.
                  1
                  2
                  3
                  4
                  5
                  6
                  7
                  8
                  9
                  10
                  11
                  12
                  Row Mean
                  SD
                  CV
                  A
                  68
                  65
                  86
                  57
                  71
                  79
                  61
                  79
                  59
                  69
                  59
                  82
                  70
                  10
                  14
                  B
                  79
                  55
                  63
                  62
                  60
                  60
                  52
                  49
                  64
                  61
                  43
                  70
                  60
                  9
                  16
                  C
                  87
                  75
                  78
                  59
                  70
                  69
                  62
                  68
                  63
                  86
                  61
                  85
                  72
                  10
                  14
                  D
                  75
                  72
                  65
                  63
                  64
                  62
                  61
                  62
                  53
                  68
                  54
                  77
                  65
                  7
                  11
                  E
                  68
                  53
                  69
                  63
                  68
                  62
                  58
                  59
                  64
                  61
                  60
                  81
                  64
                  7
                  11
                  F
                  76
                  63
                  73
                  83
                  73
                  70
                  83
                  65
                  70
                  84
                  66
                  87
                  74
                  8
                  11
                  G
                  78
                  62
                  55
                  44
                  68
                  51
                  56
                  54
                  55
                  68
                  62
                  83
                  61
                  11
                  18
                  H
                  97
                  76
                  89
                  77
                  80
                  83
                  85
                  71
                  87
                  88
                  78
                  81
                  83
                  7
                  9
                  Column Mean
                  78
                  65
                  72
                  63
                  69
                  67
                  65
                  63
                  64
                  73
                  61
                  81
                  SD
                  10
                  9
                  11
                  12
                  6
                  10
                  12
                  10
                  11
                  11
                  10
                  5
                  CV
                  12
                  13
                  16
                  19
                  8
                  16
                  19
                  15
                  17
                  15
                  16
                  6
                  Table 2. The same plate washer after preventative maintenance.
                  Assess your plate washer performance with the SMC® Plate Washer Evaluation Kit and assay precious samples with confidence.
                  Learn more about SMC® technology in our Overview of Ultrasensitive Immunoassay Technology article or find more expert advice in our SMC® Tips & Tricks article.
                  Related Products
                  Sorry, an unexpected error has occurred
                  Response not successful: Received status code 500
                  For Research Use Only. Not For Use In Diagnostic Procedures.