MilliporeSigma
HomeProtein ExpressionYeast Transformation Protocols
Yeast Transformation Protocols
Introduction
Yeasts are considered model systems for eukaryotic studies as they exhibit fast growth and have dispersed cells. Moreover, replica plating and mutant isolation of yeast cells can be done with relative ease and they have a well-defined genetic system. Most significantly, yeasts have a highly versatile DNA transformation system that can be utilized effectively for protein production.
Yeast Transformation
Catalog NumberDescriptionApplicationFeatures/Benefits
YEAST1
Yeast Transformation Kit The product contains:
  • 10 μg Control yeast plasmid DNA, pRS316
  • 100 mL PLATE buffer
  • 100 mL Transformation buffer
Suitable for transformation of any strain of yeast. Convenient, flexible and sensitive, positive transformants can be obtained with as little as 10ng of DNA; the optimum efficiency is in the 0.1-3 μg range.
  • Easy and ready-to-use
  • Requires as little as 10 ng of plasmid DNA
  • Flexibility for any strain of yeast
  • Sufficient for over 100 standard transformations
L4158Lithium acetate dihydrateFacilitate yeast transfectionPart of the BioXtra product line
T9285Tris-EDTA buffer solution TE Buffer contains 1 M Tris-HCl (pH ~8.0), containing 0.1 M EDTA.
TE (Tris EDTA) Buffer is commonly used in molecular biology labs. TE solubilizes DNA and RNA while protecting it from degradation
Suitable for storage of RNA and DNA, including purified plasmid DNA stocks
D8418Dimethyl sulfoxide (DMSO)
Can be used for buffer preparation for transformation, DEAE-dextran-mediated transfection of cells and cell fusion procedures with 40-50 % PEG; among several other applications
  • The product can increase transformation efficiency 2 to 5-fold
  • This product is free from DNases, RNases, phosphatases and proteases.
D9156Deoxyribonucleic acid (DNA), single stranded from salmon testes
Can be used for yeast transformations
This DNA is ethanol precipitated and sonicated to produce single-stranded fragments which co-migrate with the 587 and 831 base pair marker fragments
Table 1Sigma-Aldrich Yeast Transformation Products
Yeast Transformation Protocol
Protocol Overview
The LiAc transformation method involves three main steps: preparing competent yeast cells, transformation with plasmid DNA, and subsequent plating to select the transformants. Yeast transformants are usually selected using auxotrophic markers; hence, the appropriate synthetic dropout medium is used for screening transformants.
Reagents Required:
Stock Solutions
50 % Polyethylene glycol solution (mol wt ~36,500)
1 M Tris-HCl and 0.2 M EDTA, pH 8.0 (TE Buffer) (Catalog Number T9285)
1 M Lithium acetate, pH 7.5 (Catalog Number L4158)
1x TE-LiAc solution
10 mM Tris-HCl, pH 8.0, with 1 mM EDTA and 0.1 M lithium acetate
PEG-TE-LiAc solution
40 % PEG in 10 mM Tris-HCl, pH 8.0, with 1 mM EDTA and 0.1 M Lithium actetate
Yeast Transformation
  1. Prepare YPD and synthetic complete (SC) drop-out medium plates and autoclave them separately (Tables 1 and 2 ).
  2. Inoculate yeast cells from plates into 20 mL of YPD medium in a 100 mL sterile flask.
  3. Grow overnight with shaking.
  4. Dilute cells from above culture into 100 mL of YPD medium until the OD600 is 0.3
  5. Pellet cells gently.
  6. Resuspend in 7-8 mL of 1x TE-LiAc solution and rotate at 23 °C for 1-1.5 hours.
  7. Add 10 µL of 10 mg/mL salmon testes DNA (Catalog Number D9156) in sterile microfuge tubes designated for transformation and one for a negative control.
  8. Add 0.1 µg of yeast plasmid DNA (to be studied) to each tube and 100 µL of competent cells into each tube and then vortex.
  9. Add 600 µL of freshly prepared PEG-TE-LiAc solution, vortex, and incubate at 30 °C for 30 minutes with shaking.
  10. Optional - DMSO (Catalog Number D8418) can be added to 10% (v/v); followed by heat shock for 15 minutes at 42 °C.
  11. Spin for 3 seconds, resuspend cells in sterile water and plate using appropriate SC drop-out medium.
Note: Refer to growth protocols for plating yeasts and the following section for isolating transformants.
Isolation of Yeast Transformants
Yeast transformants are usually selected using the URA3 complementation method, although techniques utilizing amino acids for complementation and blue-white screening methods can also be used.
In the URA3-based selection technique; the plasmid DNA has a normal copy of the yeast URA3 gene, as well as the URA3 promoter; whereas, the yeast mutant strain lacks the URA3 allele. Thus, yeast cells transformed with the plasmid have a functional copy of the URA3 gene, which allows them to grow on Yeast Synthetic Drop-out Medium Supplements without Uracil (Catalog Number Y1501).
Figure 1.Yeast Transformation
Materials
Sorry, an unexpected error has occurred
Response not successful: Received status code 500
References
1.
Sherman F. 2002. Getting started with yeast.3-41. https://doi.org/10.1016/s0076-6879(02)50954-x
2.
MacDonald PN. 2001. Two-Hybrid Systems. https://doi.org/10.1385/1592592104
3.
Treco DA, Winston F. 2008. Growth and Manipulation of Yeast. Current Protocols in Molecular Biology. 82(1): https://doi.org/10.1002/0471142727.mb1302s82
4.
Schiestl RH, Gietz RD. 1989. High efficiency transformation of intact yeast cells using single stranded nucleic acids as a carrier. Curr Genet. 16(5-6):339-346. https://doi.org/10.1007/bf00340712
5.
Li B, Fields S. 1993. Identification of mutations in p53 that affect its binding to SV40 large T antigen by using the yeast two?hybrid system. FASEB j.. 7(10):957-963. https://doi.org/10.1096/fasebj.7.10.8344494