MilliporeSigma
HomeBiopharmaceutical CharacterizationMiddle Up Mass Analysis Using Trastuzumab as Model Protein
Biosimilar Comparability Workflow for Middle-Up Mass Analysis Using Trastuzumab as Model Protein
Geoffrey Rule1, Sundaram Palaniswamy2, Uma Sreenivasan3, Paresh Tank4, Fatima D’Souza4
1Bellefonte, PA, USA, 2Bangalore, India, 3Round Rock, TX, USA, 4ZRAS, Zelle Biotechnology Pvt. Ltd., Thane, India
Introduction
Biosimilar mAb drugs have surged in the last decade due to patent expiration of innovator drugs, offering affordable care to patients. Biosimilars, while sharing amino acid sequences with innovators, can differ structurally due to post-translational modifications (PTMs) resulting from changes in manufacturing processes. These variations, like N-glycosylation and alterations to termini, pose challenges in proving comparability.1
Thorough characterization of therapeutic mAbs is vital for safety and efficacy and establishing critical quality attributes (CQAs) are essential for both innovator and biosimilar drugs.2, 3 Characterization generally involves chromatography (e.g., SEC, RP, HILIC) combined with mass spectrometry (MS).
Trastuzumab received approval for medical usage in the United States in September 1998 and in the European Union in August 2000. At present, five biosimilars have gained approval in both the European Union and the United States.4, 5
This note describes the application of a reducing RPLC-MS workflow for middle-up comparison of trastuzumab innovator and biosimilar. The application note also includes the correlation of de-charged masses of respective heavy and light chains with theoretical masses.
A recombinant human antibody standard, SILuTMLite SigmaMAb (MSQC4), is utilized as a reference and assay control sample. Here, this is referred to simply as SigmaMAb although several other SigmaMAb standards are also available commercially.
Workflow for Middle-Up Mass Analysis of Trastuzumab
Antibody reduction (optional)
Icon for optional reduction process
    Chromatography
    Icon for chromatography
      Measurement and Analysis
      Icon for measurement and analysis
        An RPLC-MS workflow has been developed to simplify mass analysis of reduced monoclonal antibodies (mAbs) for biosimilarity assessment.
        In detail, it includes:
        • Antibody reduction procedures
        • Mass spectrometer calibration
        • System suitability test utilizing a recombinant human monoclonal antibody reference
        • An RPLC-MS method compatible with sample separation and analysis of reduced monoclonal antibodies
        General Procedures - Sample and Reference Preparation and System Setup
        Antibody Reduction Procedure
        All mAb samples were solubilized in 100 μL water at a concentration of 1 mg/mL.
        Full reduction of samples is performed by this protocol:
        1. Prepare a 100 mM solution of tris(2-carboxyethyl)phosphine (TCEP) in 6 M aqueous guanidine hydrochloride by dissolving 2.87 g of TCEP and 57.32 g of guanidine hydrochloride in 100 mL of water (if less solution is needed, scale down accordingly).
        2. Combine 30 μL of the resulting solution with 10 μL of sample.
        3. Incubate for two hours at 37 °C.
        Note: Full reduction is performed in denaturing conditions, where both the inter-chain and intra-chain disulfide bonds are broken.  For a partial reduction procedure see the application note Protocol for Purification, Optional Reduction, and SEC-MS Analysis of a Monoclonal Antibody.6
        Instrument Calibration
        The Waters QToF Xevo G2XS mass spectrometer was calibrated in a mass range of 400 – 4000 m/z with a 10 μL/min infusion of 0.4 mg/mL of sodium iodide in water. Calibration can also be performed with a 10 μL/min infusion of 0.4 mg/mL cesium iodide and polyalanine in water, prior to running the samples.
        System Suitability
        Reduced SigmaMAb antibody reference (formulated at 2 mg/mL and further diluted to 1 mg/mL) was analyzed alongside samples to determine system suitability.
        RPLC-MS System Setup and MS Data Analysis
        RPLC-MS System Setup
        Essential settings of the UHPLC-PDA chromatography system and qToF mass spectrometer are listed in Tables 1 and 2 below.
        LC  Conditions
        Instrument:
        WatersTM H-Class Acquity UPLC Chromatography System
        Software:
        UNIFI
        Column:
        BIOshell™ A400 Protein C4, 100 x 2.1 mm I.D., 3.4 μm (66825-U)
        Mobile phase:
        [A] Water + 0.1% FA; [B] acetonitrile + 0.1% FA
        Gradient:
        See Table 1b
        Flow:
        0.4 mL/min
        Column temp:
        80 °C
        Autosampler temp:
        8 °C
        Run time:
        20 min
        Detector:
        PDA 280 nm and MS TOF (see Table 2 for conditions)
        Flow divert:
        1.0 – 20.0 min
        Loop volume:
        10 μL
        Injection volume:
        2 μL
        Table 1.UHPLC-PDA settings
        Time (min)
        %A
        %B
        Curve
        0.00
        75.0
        25.0
        Initial
        1.00
        75.0
        25.0
        6
        8.00
        65.0
        35.0
        6
        11.00
        40.0
        60.0
        6
        14.00
        5.0
        95.0
        6
        14.90
        5.0
        95.0
        6
        15.00
        75.0
        25.0
        6
        20.00
        75.0
        25.0
        6
        Table 1b.Gradient table for reduced mass analysis
        MS Conditions
        Instrument:
        WatersTM QToF Xevo G2X2 Mass Spectrometer
        Software:
        UNIFI
        Capillary (V):
        3,000
        Sample cone (V)
        40
        Extraction cone (V):
        3
        Ion guide (V):
        3
        Desolvation temp (° C):
        100
        Source temp (° C):
        120
        Scan range (Da):
        400 – 4,000
        Desolvation gas (L/h):
        800
        Cone gas (L/h):
        50
        Collision energy (V):
        5
        Pusher (V):
        930
        RF Setting:
        Autoprofile
        Table 2.qToF-MS settings
        MS Data Analysis
        Data were processed using the MaxEnt1 module within the UNIFI software to generate deconvoluted mass spectra. In general, a summed spectrum was created from the total ion chromatogram (TIC) of the heavy chain (HC), or light chain (LC), subunits and the spectrum then processed by MaxEnt1; detailed parameters in Table 3. Glycoforms were matched by the software and HC glycans are listed in the Results section. Relative abundance data were tabulated based on peak intensities of the coeluting glycoform species. 
        Reduced
        m/z Range:
        750 – 2,250
        Mass range (Da):
        LC 20,000 - 25,000; HC 45,000 - 55,000
        Output resolution:
        1 Da
        Peak width model:
        tof
        TOF Resolution:
        10,000
        Minimum intensity ratios, L and R (%) :
        30
        Iterations:
        21
        Table 3.Deconvolution parameters
        Results
        Intact Mass Analysis of Reduced Trastuzumab
        The objective of this analysis was to compare the molecular weight of trastuzumab innovator and biosimilar, light, and heavy chains, using an RPLC-MS, middle-up approach.
        System Suitability Test Results
        Four microliters of reduced SigmaMAbTM reference sample was injected onto the RPLC-MS system. The observed heavy chain glycoforms matched the expected glycoform masses of SigmaMAbTM, as listed in Table 4. Discrepancies between the observed masses and the theoretical values for four glycoforms are all within 0.003% mass error. Figure 1 illustrates the UV (280 nm) and TIC chromatograms of the SigmaMAbTM reference, while Figure 2 shows deconvoluted mass spectra of the light and heavy chain glycoforms.
        A UV chromatogram of reduced SigmaMAb reference standard, measured at 280 nm, displays two distinct peaks at 5.87 and 7.55 minutes, with absorbance on the y-axis and retention time on the x-axis.
        A TIC chromatogram of reduced SigmaMAb reference standard displays two distinct peaks at 5.95 and 7.63 minutes, with total ion current on the y-axis and retention time on the x-axis.
        Figure 1. Photodiode array (280 nm, left) and TIC trace (right) of reduced SigmaMAbTM reference.
        Deconvoluted mass spectra obtained for light chain of fully reduced SigmaMAb reference with a distinct peak at an observed mass of 22942
        Deconvoluted mass spectra of the heavy chain from fully reduced SigmaMAb reference reveal three distinct peaks observed at mass values of 50403, 50565, and 50727 respectively, from left to right.
        Figure 2. Deconvoluted mass spectra of light and heavy chains (left and right, respectively) of fully reduced SigmaMAbTM reference.
        Species
        Molecular Formula
        Theoretical Mass (Da) Fully Reduced*
        Intra Disulfide Bonds
        Theoretical Mass (Da) Partially Reduced*
        Observed Mass (Da)
        Error (%)
        Light chain
        C1006H1555N267O333S7
        22,942.2
        2 (-4 Da)
        22,938.2
        22942
        -0.001
        Heavy Chain/G0F
        C2237H3485N591O702S16
        50,403.2
        4 (-8 Da)
        50,395.2
        50403
        -0.000
        Heavy Chain/G1F
        C2243H3495N591O707S16
        50,565.3
        4 (-8 Da)
        50,557.3
        50565
        -0.001
        Heavy Chain/G2F
        C2249H3505N591O712S16
        50,727.5
        4 (-8 Da)
        50,719.5
        50726
        -0.003
        G0F: GlcNAc2Man3GlcNAc2Fuc
        G1F: GalGlcNAc2Man3GlcNAc2Fuc
        G2F: Gal2GlcNAc2Man3GlcNAc2Fuc
        *Masses based on NIST Physical Reference Data
        Table 4.Theoretical and observed masses of reduced SigmaMAbTM reference light and heavy chain glycoforms
        Reduced Sample Results
        The fully reduced trastuzumab innovator and biosimilar samples were also analyzed using RPLC-MS. The corresponding photodiode array (280 nm) traces, TICs, and the summed and deconvoluted MS spectra of the samples are shown in Figures 3 and 4. The measured masses of reduced forms correlated well with the calculated masses as shown in Table 5 (mass error 0.001%, or less, for innovator and 0.003%, or less, for biosimilar). 
        Figure 3. Comparative photodiode array (280 nm, left) and TIC trace (right) of fully reduced trastuzumab. Red: Innovator; Blue: Biosimilar.
        Summed up mass spectra for light chain of fully reduced trastuzumab. The innovator is represented by a red curve, while the biosimilar is depicted by a blue curve, with intensity measured in counts on the y-axis and observed mass on the x-axis.
        Summed up mass spectra for heavy chain of fully reduced trastuzumab. The innovator is represented by a red curve, while the biosimilar is depicted by a blue curve, with intensity measured in counts on the y-axis and observed mass on the x-axis.
        Deconvulated mass spectra for light chain of fully reduced trastuzumab with a distinct peak observed at a mass of 23443. The innovator is represented by a red curve, while the biosimilar is depicted by a blue curve, with intensity measured in counts on the y-axis and observed mass on the x-axis.
        The deconvoluted mass spectra for the heavy chain of fully reduced trastuzumab reveal distinct peaks at mass values of 50601, 50763, and 50925 from left to right. The innovator is depicted by a red curve, and the biosimilar by a blue curve, with intensity measured in counts on the y-axis and observed mass on the x-axis.
        Figure 4. Comparative summed (top) and deconvoluted (bottom) mass spectra of light and heavy chains (left and right, respectively) of fully reduced trastuzumab. Red: Innovator; Blue: Biosimilar.
        Species
        Theoretical Mass (Da) Fully Reduced*
        Intra Disulfide Bonds
        Theoretical Mass (Da) Partially Reduced*
        Observed Mass (Da)
        Error (%)
        Innovator
        Biosimilar
        Innovator
        Biosimilar
        Light chain
        23442.8
        2 (-4 Da)
        23,438.8
        23443
        23443
        +0.001
        +0.001
        Heavy chain/G0F
        50601.2
        4 (-8 Da)
        50,593.2
        50601
        50601
        +0.000
        +0.000
        Heavy chain/G1F
        50763.4
        4 (-8 Da)
        50,755.4
        50763
        50763
        -0.001
        -0.001
        Heavy chain/G2F
        50925.5
        4 (-8 Da)
        50,917.5
        50925
        50927
        -0.001
        +0.003
        G0F: GlcNAc2Man3GlcNAc2Fuc
        G1F: GalGlcNAc2Man3GlcNAc2Fuc
        G2F: Gal2GlcNAc2Man3GlcNAc2Fuc
        Table 5.Theoretical and observed masses of reduced trastuzumab light chain and heavy chain glycoforms (*theoretical mass calculated based on NIST Physical Reference Data)
        Conclusion
        Ensuring regulatory approvals of biosimilar mAb drug products requires meticulous comparability assessment to guarantee safety and efficacy. This application note utilizes reversed-phase liquid chromatography-mass spectrometry (RPLC-MS) at the subunit level for assessing comparability between originator trastuzumab and its biosimilar counterpart. An RP method suitable for reduced mAbs was established using a wide-pore, superficially porous C4 column.
        The workflow uses a mAb reduction procedure, performed with full reduction so that all disulfide bonds are broken. Solutions of sodium iodide, or cesium iodide, were used for mass spectrometer calibration.
        Upon analyzing the reduced SigmaMAbTM reference sample, observed glycoform masses of the heavy chain were consistent with anticipated masses with the LC and three HC glycoforms all within a 0.003% mass error threshold. Similarly, the measured masses of reduced innovator trastuzumab, and biosimilar, exhibited close alignment with theoretical masses (mass error of 0.003%, or below).
        These findings affirm the versatility of the RPLC-MS workflow for analyzing reduced monoclonal antibody samples, delivering accurate results for distinct identification of multiple glycoforms.
        See more applications on the Biopharmaceutical Characterization page.
        Related Products
        Sorry, an unexpected error has occurred
        Response not successful: Received status code 500
        References
        1.
        Review and Approval. 2022 Dec 13 . [Internet]. U.S. Food and Drug Administration. Available from: https://www.fda.gov/drugs/biosimilars/review-and-approval
        2.
        Sandra K, Vandenheede I, Sandra P. 2014. Modern chromatographic and mass spectrometric techniques for protein biopharmaceutical characterization. Journal of Chromatography A. 133581-103. https://doi.org/10.1016/j.chroma.2013.11.057
        3.
        Berkowitz SA, Engen JR, Mazzeo JR, Jones GB. 2012. Analytical tools for characterizing biopharmaceuticals and the implications for biosimilars. Nat Rev Drug Discov. 11(7):527-540. https://doi.org/10.1038/nrd3746
        4.
        Biosimilars approved in the US. Gabionline.net. [Internet]. Available from: https://www.gabionline.net/biosimilars/general/biosimilars-approved-in-the-us
        5.
        Biosimilars approved in Europe. Gabionline.net. [Internet]. Available from: https://www.gabionline.net/biosimilars/general/biosimilars-approved-in-europe
        6.
        Protocol for purification, optional reduction, and SEC-MS analysis of a monoclonal antibody. [Internet]. Sigmaaldrich.com. Available from: /US/en/technical-documents/protocol/pharmaceutical-and-biopharmaceutical-manufacturing/biologics-biosimilars-characterization/protocol-for-analysis-of-a-monoclonal-antibody