MilliporeSigma
HomeSmall Molecules Analysis & QCAssay of Doxepin Hydrochloride USP Monograph
Assay of Doxepin Hydrochloride acc. to USP Monograph
Lee May May, Application Chemist
Abstract
This study describes a rugged high-performance liquid chromatography (HPLC) method for the assay of Doxepin Hydrochloride, following monograph guidelines as specified by the United States Pharmacopeia (USP). The method is proven to be highly stable with good reproducibility and excellent peak shapes using the fully porous particle (FPP) Purospher® STAR RP-8e column over a wide concentration range of 0.01 to 1.0 mg/mL. The method complies with all USP specifications in terms of resolution, tailing factors, and RSD for both (E)- and (Z)-isomers.
Section Overview:
The chemical structure of a compound with a large ring system composed of three fused rings: two benzene rings and one other cyclic structure. Attached to this ring system is a side chain with a tertiary amine group, featuring two methyl groups (CH3) attached to the nitrogen (N). Additionally, a hydrogen chloride (HCl) molecule is depicted separately, indicating the compound is likely presented as a hydrochloride salt.
Introduction
Doxepin hydrochloride is a dibenzoxepin tricyclic compound used as an anti-depressant. The molecular formula of the compound is C19H21NO•HCl and its molecular weight is 315.84 Da. Doxepin HCl is made up of (E) and (Z) geometric isomers. It is a white crystalline powder freely soluble in water, methanol and dichloromethane.1
The assay described below follows that of the USP monograph.2 It uses the fully porous particle (FPP) Purospher® STAR RP-8e column (USP code L7) for an extended doxepin concentration range from 0.01 to 1.0 mg/mL. 
Experimental
HPLC Parameters
Column:
Purospher® STAR RP-8 endcapped Hibar® RT 125 x 4 mm I.D., 5 µm (1.50033)
Mobile Phase:
[A] Buffer: 27.6 g/L monobasic sodium phosphate (NaH2PO4);
[B] Methanol;
A:B 70:30, adjusted to pH 2.5 with phosphoric acid; isocratic
Flow rate:
1 mL/min
Pressure Drop:
185 bar
Column temp.:
50 °C
Detector:
UV at 254 nm
Injection:
20 µL
Standard:
0.1 mg/mL USP Doxepin HCl RS in mobile phase
Table 1.LC conditions used for the doxepin hydrochloride determination
Results
System Suitability
USP monograph suitability requirements
  • Run time: NLT 2 times the retention time of the (E) isomer
  • Resolution: NLT 1.5 between the (E) and (Z) isomers.
  • Tailing factor: NMT 2.0 for each (E) and (Z) isomers.
  • Relative standard deviation: NMT 2.0% each of the (E)- and (z)-isomers
Acceptance criteria
  • (E)-isomer: NLT 81.4% and NMT 88.2%.
  • (Z)-isomer: NLT 13.6% and NMT 18.1%.
Peak No.
Analyte
RT (min)
Area (mAU*min)
% Peak area
Tailing factor
Resolution
1
(E)-isomer
22.55
31.917
84.93
1.00
2
(Z)-isomer
25.74
5.665
15.07
1.02
1.84
USP Requirement
NMT 2.0
NLT 1.5
Table 2.Injection of 0.1 mg/mL doxepin HCl – Identification and system suitability
A chromatogram obtained on the HPLC-UV analysis of 0.1 mg/mL of doxepin HCl, showing absorbance in milli-absorbance units (mAU) versus time in minutes. The graph features a prominent sharp peak, labeled 1, reaching just below 35 mAU at 22.55 minutes for the (E)-isomer, followed by a smaller peak at 25.74 minutes, labeled 2, for (Z)-isomer, and a baseline close to zero throughout the rest of the timeline.
Figure 1.Analysis of 0.1 mg/mL of doxepin HCl in mobile phase. The run time is NLT 2 times the retention time of the (E) isomer (here 22.55 min) following the USP monograph specification.
(E)-Isomer
(Z)-Isomer
Injection
Peak Area
(mAU*min)
RT
 (min)
Peak Area
(mAU*min)
RT
(min)
1st
31.917
22.55
5.665
25.74
2nd
31.822
22.53
5.666
25.72
3rd
31.404
22.55
5.571
25.73
4th
31.319
22.58
5.563
25.75
5th
30.763
22.58
5.474
25.77
Mean
31.445
22.55
5.588
25.74
Standard Deviation
0.4603
0.02
0.0805
0.02
% RSD
1.47
0.10
1.44
0.08
USP Requirement (RSD)
NMT 2.0%
NMT 2.0%
Table 3.Repeatability of doxepin HCl standard injections (0.1 mg/mL) - Peak areas and retention times
Linearity & Sensitivity
The linearity of (E)- and (Z)-isomers was demonstrated for 8 concentrations from 0.01 to 1.0 mg/mL with an R2 of 0.9999 (Tables 3 & 4, Figures 2 & 3). This is 10 to 1000% of the doxepin test concentration at 0.1 mg/mL. The compendial guideline for the establishment of an assay linearity is 5 concentrations from 80% to 120% of the test concentration. 
Conc.* mg/mL
Mean Area (mAU*min)
0.0103
3.247
0.0256
8.237
0.0513
16.054
0.1026
31.613
0.2050
64.147
0.4102
127.352
0.8204
250.354
1.0255
315.945
STEYX
0.997
Slope
306.75
R2
0.9999
LOD* (mg/mL)
0.011
LOQ* (mg/mL)
0.033
*related to the used conc. of doxepin HCl RS
Table 4.Linearity and sensitivity (E)-isomer – Average peak areas of duplicates (0.01 to 1.0 mg/mL doxepin HCL RS injections)
The graph plots ‘Area (mAU*min)’ on the y-axis (ranging from 0 to 350) against ‘Concentration, mg/ml’ on the x-axis (ranging from 0 to 1.2). Eight data points (represented as blue dots) obtained for (E)-isomer using doxepin HCL RS injections at concentrations between 0.01 to 1.0 mg/mL, form an almost perfect straight line, indicating a strong linear relationship between the area and concentration of the (E)-isomer. The line of best fit is represented by the equation ‘y = 306.75x + 0.4753,’ with a coefficient of determination (R²) of 0.9999, suggesting an extremely high level of correlation between the variables.
Figure 2.Calibration curve for (E)-isomer from 0.01 to 1.0 mg/mL doxepin HCL RS injections.
Conc.* mg/mL
Mean Area (mAU*min)
0.0103
0.577
0.0256
1.433
0.0513
2.832
0.1026
5.619
0.2050
11.418
0.4102
22.771
0.8204
44.825
1.0255
56.552
STEYX
0.167
Slope
54.951
R2
0.9999
LOD* (mg/mL)
0.010
LOQ* (mg/mL)
0.030
*Related to the used conc. of doxepin HCl RS
Table 5. Linearity and sensitivity for (Z)-isomer – Average peak area of duplicates (0.01 to 1.0 mg/mL doxepin HCL RS injections)
The graph plots ‘Area (mAU*min)’ on the y-axis (ranging from 0 to 70) against ‘Concentration, mg/ml’ on the x-axis (ranging from 0 to 1.2). Eight data points obtained for (Z)-isomer using doxepin HCL RS injections at concentrations between 0.01 to 1.0 mg/mL, form an almost perfect straight line, indicating a strong linear relationship between the area and concentration of the (E)-isomer. The line of best fit is represented by the equation ‘y = 54.951x + 0.045,’ with a coefficient of determination (R²) of 0.9999, suggesting an extremely high level of correlation between the variables.
Figure 3.Calibration curve for (Z)-isomer from 0.01 to 1.0 mg/mL doxepin HCL RS injections.
Conclusion
The Purospher® STAR RP-8e column was able to comply with the criteria set out for the assay of doxepin HCl in the USP monograph and demonstrated a solid performance:
  • The resolution between (E)- and (Z)-isomers was 1.84. (criteria NLT 1.5)
  • The peak symmetry factor of the doxepin (E)-isomer ranged from 0.96 to 1.32 for a concentration from 0.01 to 1 mg/mL. (criteria NMT 2.0)
  • % RSD for 5 repeats at 0.1 mg/mL doxepin was 1.47 and 1.44 for the peak areas of the (E)- and (Z)-isomers respectively. (criteria NMT 2%)
  • The retention time of (E)- and (Z)-isomers was 22.55 and 25.74 min respectively. (RSD < 2% for n=5)
  • LOD and LOQ of the doxepin (E)-isomer was 0.011 and 0.033 mg/mL respectively.*
  • LOD and LOQ of the doxepin (Z)-isomer was 0.010 and 0.030 mg/mL respectively.*
*related to the used conc. of doxepin HCl RS
The column was robust and worked well at the low pH of 2.5 at an elevated temperature of 50 °C. Extended repeated injections (data not shown) showed good reproducibility for the retention times so long as the pH is well controlled within ±0.1 units. The column provided excellent peak shapes and was linear for an extended test range – both of which is essential for accurate quantification. 
See more applications for Pharmaceutical Analysis and Quality Control
Related Products
Sorry, an unexpected error has occurred
Response not successful: Received status code 500
References
1.
PubChem. Doxepin. Nih.gov. . [Internet]. Available from: https://pubchem.ncbi.nlm.nih.gov/compound/Doxepin
2.
United States Pharmacopeia (2021). USP Monographs, Doxepin Hydrochloride. [Internet]. Rockville, MD: United States Pharmacopeia.[cited 11 Apr 2022]. Available from: https://doi.usp.org/USPNF/USPNF_M28110_07_01