Merck
New plasmid-mediated fluoroquinolone efflux pump, QepA, found in an Escherichia coli clinical isolate.
Antimicrobial agents and chemotherapy (2007-06-06)
Kunikazu Yamane, Jun-Ichi Wachino, Satowa Suzuki, Kouji Kimura, Naohiro Shibata, Haru Kato, Keigo Shibayama, Toshifumi Konda, Yoshichika Arakawa
ABSTRAKT
Plasmid-mediated Qnr and AAC(6')-Ib-cr have been recognized as new molecular mechanisms affecting fluoroquinolone (FQ) resistance. C316, an Escherichia coli strain demonstrating resistance to various FQs, was isolated in Japan. Resistance to FQs was augmented in an E. coli CSH2 transconjugant, but PCR failed to detect qnr genes, suggesting the presence of novel plasmid-mediated FQ resistance mechanisms. Susceptibility tests, DNA manipulation, and analyses of the gene and its product were performed to characterize the genetic determinant. A novel FQ-resistant gene, qepA, was identified in a plasmid, pHPA, of E. coli C316, and both qepA and rmtB genes were mediated by a probable transposable element flanked by two copies of IS26. Levels of resistance to norfloxacin, ciprofloxacin, and enrofloxacin were significantly elevated in E. coli transformants harboring qepA under AcrB-TolC-deficient conditions. QepA showed considerable similarities to transporters belonging to the 14-transmembrane-segment family of environmental actinomycetes. The effect of carbonyl cyanide m-chlorophenylhydrazone (CCCP) on accumulation of norfloxacin was assayed in a qepA-harboring E. coli transformant. The intracellular accumulation of norfloxacin was decreased in a qepA-expressing E. coli transformant, but this phenomenon was canceled by CCCP. The augmented FQ resistance level acquired by the probable intergeneric transfer of a gene encoding a major facilitator superfamily-type efflux pump from some environmental microbes to E. coli was first identified. Surveillance of the qepA-harboring clinical isolates should be encouraged to minimize further dissemination of the kind of plasmid-dependent FQ resistance determinants among pathogenic microbes.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Erythromycin, meets USP testing specifications
Sigma-Aldrich
Tetracycline, 98.0-102.0% (HPLC)
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Erythromycin, tested according to Ph. Eur.
Sigma-Aldrich
Ethidium bromide solution, suitable for fluorescence, ~1% in H2O
Sigma-Aldrich
Erythromycin, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Rhodamine 6G, Dye content ~95 %
Sigma-Aldrich
Erythromycin, potency: ≥850 μg per mg
Sigma-Aldrich
Ethidium bromide solution, BioReagent, Molecular Biology, 500 μg/mL in H2O
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, Molecular Biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Rhodamine 6G, Dye content 99 %
Sigma-Aldrich
Gentian Violet, meets USP testing specifications
Sigma-Aldrich
Deoxycholic acid, ≥98% (HPLC)
Sigma-Aldrich
Crystal Violet, certified by the BSC
Sigma-Aldrich
Crystal Violet, ACS reagent, ≥90.0% anhydrous basis
Sigma-Aldrich
Crystal Violet, suitable for microscopy
Sigma-Aldrich
Ciprofloxacin, ≥98% (HPLC)
Supelco
Ciprofloxacin, VETRANAL®, analytical standard
Supelco
Enrofloxacin, VETRANAL®, analytical standard
Sigma-Aldrich
Ethidium bromide, ~95% (HPLC)
Sigma-Aldrich
Ethidium bromide, BioReagent, Molecular Biology, powder