Merck
Tumor necrosis factor reduces the amplitude of rat cortical spreading depression in vivo.
Annals of neurology (2014-05-07)
Frank Richter, Wibke Lütz, Annett Eitner, Johannes Leuchtweis, Alfred Lehmenkühler, Hans-Georg Schaible
ABSTRAKT
Brain damage and ischemia often trigger cortical spreading depression (CSD), which aggravates brain damage. The proinflammatory cytokine tumor necrosis factor (TNF) is significantly upregulated during brain damage, but it is unknown whether TNF influences spreading depression in cerebral cortex in vivo. This question is important because TNF not only furthers inflammatory reactions but might also be neuroprotective. Here we tested the hypothesis that TNF affects CSD, and we explored the direction in which CSD is modified by TNF. CSD, elicited by pressure microinjection of KCl, was recorded in anesthetized rats and mice. TNF was administered locally into a trough, providing local TNF treatment of a cortical area. For further analysis, antibodies to TNF receptor (TNFR) 1 or 2 were applied, or CSD was monitored in TNFR1 and TNFR2 knockout mice. γ-Aminobutyric acid (GABA)A receptors were blocked by bicuculline. Immunohistochemistry localized the cortical expression of TNFR1 and TNFR2. Local application of TNF to the cortex reduced dose-dependently the amplitude of CSD. This effect was prevented by blockade or knockout of TNFR2 but not by blockade or knockout of TNFR1. TNFR2 was localized at cortical neurons including parvalbumin-positive inhibitory interneurons, and blockade of GABAA receptors by bicuculline prevented the reduction of CSD amplitudes by TNF. We identified a functional link between TNF and CSD. TNF activates TNFR2 in cortical inhibitory interneurons. The resulting release of GABA reduces CSD amplitudes. In this manner, TNF might be neuroprotective in pathological conditions.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
γ-Aminobutyric acid, ≥99%
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
SAFC
Sodium chloride solution, 5 M
Supelco
γ-Aminobutyric acid, analytical standard
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
γ-Aminobutyric acid, BioXtra, ≥99%
Sigma-Aldrich
Sodium chloride, BioUltra, Molecular Biology, ≥99.5% (AT)
Vigabatrin impurity D, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Urethane, ≥99%
Sigma-Aldrich
Urethane, ≥99.0% (GC)
Supelco
Urethane, analytical standard
Sigma-Aldrich
Calcium chloride
Sigma-Aldrich
Calcium chloride, anhydrous, BioReagent, suitable for insect cell culture, suitable for plant cell culture, ≥96.0%
Supelco
Calcium ion solution for ISE, 0.1 M Ca, analytical standard (for ion-selective electrodes)