Merck
Stearidonic acid-enriched flax oil reduces the growth of human breast cancer in vitro and in vivo.
Breast cancer research and treatment (2014-11-25)
K Subedi, H-M Yu, M Newell, R J Weselake, D Meesapyodsuk, X Qiu, S Shah, C J Field
ABSTRAKT
The 20 and 22 carbon n-3 long-chain polyunsaturated fatty acids (LCPUFA) inhibit the growth of tumors in vitro and in animal models, but less is known about the 18 carbon n-3, stearidonic acid (SDA). This study aimed to establish and determine a mechanism for the anti-cancer activity of SDA-enriched oil (SO). SO (26 % of lipid) was produced by genetically engineering flax and used to treat human tumorigenic (MDA-MB-231, MCF-7) and non-tumorigenic (MCF-12A) breast cells. Nu/nu mice bearing MDA-MB-231 tumor were fed SO (SDA, 4 % of fat). Cell/tumor growth, phospholipid (PL) composition, apoptosis, CD95, and pro-apoptotic molecules were determined in SO-treated cells/tumors. Compared to a control lipid mixture, SO reduced (p < 0.05) the number of tumorigenic, but not MCF-12A cells, and resulted in higher concentration of most of the n-3 fatty acids in PL of all cells (p < 0.05). However, docosapentaenoic acid increased only in tumorigenic cells (p < 0.05). SO diet decreased tumor growth and resulted in more n-3 LCPUFA, including DPA and less arachidonic acid (AA) levels in major tumor PL (p < 0.05). Treatment of MDA-MB-231 cells/tumors with SO resulted in more apoptotic cells (in tumors) and in vivo and in vitro, more CD95+ positive cells and a higher expression of apoptotic molecules caspase-10, Bad, or Bid (p < 0.05). Supplementing SO alters total PL and PL classes by increasing membrane content of n-3 LCPUFA and lowering AA (in vivo), which is associated with increased CD95-mediated apoptosis, thereby suggesting a possible mechanism for reduce tumor survival.
MATERIAŁY
Numer produktu
Marka
Opis produktu
USP
Palmitic acid, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Palmitic acid, natural, 98%, FG
Palmitic acid, European Pharmacopoeia (EP) Reference Standard
Supelco
Palmitic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Linoleic acid, technical, 58-74% (GC)
Sigma-Aldrich
5-Aminolevulinic acid hydrochloride, ≥97.0% (AT)
Sigma-Aldrich
Trypan Blue, ≥80% (HPLC), Dye content 60 %
Sigma-Aldrich
Palmitic acid, ≥98% palmitic acid basis (GC)
Sigma-Aldrich
Palmitic acid, BioXtra, ≥99%
Sigma-Aldrich
Palmitic acid, ≥98%, FCC, FG
Sigma-Aldrich
5-Aminolevulinic acid hydrochloride, ≥98%
Sigma-Aldrich
5-Aminolevulinic acid hydrochloride, BioReagent, suitable for cell culture, powder, ≥98%
Sigma-Aldrich
Linolenic acid, ≥99%
Sigma-Aldrich
Trypan Blue, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Palmitic acid, ≥99%
Sigma-Aldrich
Linolenic acid, ~70% (GC)
Sigma-Aldrich
Linoleic acid, liquid, BioReagent, suitable for cell culture
Supelco
Palmitic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Linolenic acid, analytical standard
Supelco
Linoleic acid, analytical standard
Supelco
Palmitic acid, analytical standard
Sigma-Aldrich
Stearic acid, ≥95%, FCC, FG
Sigma-Aldrich
Stearic acid, reagent grade, 95%
Supelco
Stearic Acid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Oleic acid, ≥99% (GC)
Sigma-Aldrich
Oleic acid, suitable for cell culture, BioReagent
Sigma-Aldrich
Trypan Blue solution, 0.4%, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Stearic acid, Grade I, ≥98.5% (capillary GC)
Sigma-Aldrich
β-D-Allose, rare aldohexose sugar
Sigma-Aldrich
α-Linoleic acid, ≥98%