Merck
  • Antibody ligation of CM1 on cisplatin-exposed HeLa cells induces apoptosis through reactive oxygen species-dependent Fas ligand expression.
Antibody ligation of CM1 on cisplatin-exposed HeLa cells induces apoptosis through reactive oxygen species-dependent Fas ligand expression.
International journal of oncology (2014-04-01)
Ga Bin Park, Daejin Kim, Hoi Soo Yoon, Yeong-Seok Kim, Hyun-Kyung Lee, Ki Tae Kim, Dae Hoon Jeong, Dae Young Hur
ABSTRAKT
Centrocyte/centroblast marker 1 (CM1) has been identified as a pro-apoptosis molecule on B-cell lymphoma cells as well as several types of cancer cells. In this study, we investigated its signaling mechanism in HeLa cells after treatment with cisplatin in order to potentially identify a new therapeutic target. The CM1 molecule was induced on the surface of cisplatin-exposed HeLa cells. In these cells, ligation of CM1 with anti-CM1 monoclonal antibodies inhibited cell proliferation and produced reactive oxygen species. Fas ligand (FasL) expression was upregulated without upregulating Fas in cisplatin-exposed HeLa cells after CM1 stimulation. Pretreatment with N-acetylcysteine, a pan-capase inhibitor, and ZB4, an antagonistic anti-Fas antibody, effectively inhibited the apoptotic effect triggered by CM1. CM1 ligation induced apoptosis through disruption of the mitochondrial membrane potential, decreased Bcl-2 and phosphorylated ERK expression. These findings identify CM1 as a potential new therapeutic target related to cisplatin-exposed cervical cancer.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, suitable for protein labeling, ≥90% (HPLC), powder
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Sigma-Aldrich
cis-Diamineplatinum(II) dichloride, ≥99.9% trace metals basis
USP
Transplatyna, United States Pharmacopeia (USP) Reference Standard
Acetylcysteine, European Pharmacopoeia (EP) Reference Standard
USP
Acetylocysteina, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Dioctan 2′,7′-dichlorodihydrofluoresceiny, ≥97%
Cisplatin impurity A, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
4-(2-Aminoethyl)benzenesulfonyl fluoride hydrochloride, ≥97.0% (HPLC)
Sigma-Aldrich
trans-Platinum(II)diammine dichloride
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, ≥90% (HPLC)
Supelco
Pefabloc® SC
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, BioReagent, suitable for fluorescence, mixture of 2 components, ≥90% (HPLC)
Cisplatin, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
cis-Diammineplatinum(II) dichloride, crystalline
Supelco
N-Acetyl-L-cysteine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
N-Acetyl-L-cysteine, BioXtra, ≥99% (TLC)
Sigma-Aldrich
N-Acetyl-L-cysteine, Sigma Grade, ≥99% (TLC), powder
Sigma-Aldrich
N-Acetyl-L-cysteine, BioReagent, suitable for cell culture