Merck
Simultaneous quantification of seven hippocampal neurotransmitters in depression mice by LC-MS/MS.
Journal of neuroscience methods (2014-04-17)
Fei Huang, Jia Li, Hai-Lian Shi, Ting-ting Wang, Wahaf Muhtar, Min Du, Bei-bei Zhang, Hui Wu, Li Yang, Zhi-bi Hu, Xiao-jun Wu
ABSTRAKT
There is no method available to simultaneously detect GABA, Glu, Epi, NE, DA, 5-HT and 5-HIAA in mouse hippocampus. A rapid and sensitive LC-MS/MS method has been developed for simultaneously measuring seven neurotransmitters in mouse hippocampus. The analytes were detected in positive mode with multiple reaction monitoring (MRM) and the procedure was completed in less than 9min. This method exhibited excellent linearity for all of the analytes with regression coefficients higher than 0.99, and showed good intra- and inter-day precisions (RSD<15%) with good accuracy (80-120%). Moreover, the method was successfully applied for the quantitative determination of neurotransmitters in a mouse depression model induced by successive methylprednisolone injections. The results indicated that this depression model was closely associated with the decreased level of Epi (p=0.002) and elevated ratio of 5-HIAA/5-HT (p=0.01), which has never been reported elsewhere. Compared with previous methods, current approach is more convenient without any pre-column derivatization of the analytes but enhances detectability with incremental neurotransmitter profile and shortens detection time. This work represents the first accurate simultaneous determination of seven neurotransmitters in the mouse depression model induced by methylprednisolone. The reliable method will benefit the research of neurological diseases with the altered neurotransmitter profile in brain.
MATERIAŁY
Numer produktu
Marka
Opis produktu
USP
Metyloprednizolon, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Serotonin creatinine sulfate monohydrate, powder
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Methylprednisolone, European Pharmacopoeia (EP) Reference Standard
USP
Rozpuszczalnik resztkowy klasy 2 - acetonitryl, United States Pharmacopeia (USP) Reference Standard
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
6α-Methylprednisolone, ≥98%
Sigma-Aldrich
Formic acid, reagent grade, ≥95%
Sigma-Aldrich
Formic acid solution, BioUltra, 1.0 M in H2O
Sigma-Aldrich
Formic acid, ACS reagent, ≥96%
Sigma-Aldrich
Formic acid, ACS reagent, ≥88%
USP
Alkohol metylowy, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Glutamic acid, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%