Merck
Computational and functional characterization of Angiogenin mutations, and correlation with amyotrophic lateral sclerosis.
PloS one (2014-11-06)
Aditya K Padhi, Kamalika Banerjee, James Gomes, Manidipa Banerjee
ABSTRAKT
The Angiogenin (ANG) gene is frequently mutated in patients suffering from the neurodegenerative disease--amyotrophic lateral sclerosis (ALS). Most of the ALS-causing mutations in Angiogenin affect either its ribonucleolytic or nuclear translocation activity. Here we report the functional characterization of two previously uncharacterized missense mutations in Angiogenin--D22G and L35P. We predict the nature of loss-of-function(s) in these mutants through our previously established Molecular Dynamics (MD) simulation extended to 100 ns, and show that the predictions are entirely validated through biochemical studies with wild-type and mutated proteins. Based on our studies, we provide a biological explanation for the loss-of-function of D22G-Angiogenin leading to ALS, and suggest that the L35P-Angiogenin mutation would probably cause ALS symptoms in individuals harboring this mutation. Our study thus highlights the strength of MD simulation-based predictions, and suggests that this method can be used for correlating mutations in Angiogenin or other effector proteins with ALS symptoms.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Sodium chloride solution, 0.85%
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Streptomycin sulfate salt, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Streptomycin sulfate salt, powder
Sigma-Aldrich
Streptomycin sulfate salt, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, BioUltra, Molecular Biology, ≥99.5% (AT)
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Streptomycin sulfate, European Pharmacopoeia (EP) Reference Standard
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Perchloric acid, 0.01 M HClO4 in water (0.01N), eluent for IC
Sigma-Aldrich
Perchloric acid, 70%, 99.999% trace metals basis
Supelco
Streptomycin solution, ~1 mg/mL in 1 mM EDTA, analytical standard
Sigma-Aldrich
Streptomycin, Ready Made Solution, 100 mg/mL in water
Sigma-Aldrich
Perchloric acid, ACS reagent, 60%
Sigma-Aldrich
Perchloric acid, ACS reagent, 70%