Merck
IL-18 synergizes with IL-7 to drive slow proliferation of naive CD8 T cells by costimulating self-peptide-mediated TCR signals.
Journal of immunology (Baltimore, Md. : 1950) (2014-09-10)
Matthew C Walsh, Erika L Pearce, Pedro J Cejas, JangEun Lee, Li-San Wang, Yongwon Choi
ABSTRAKT
Naive T cell populations are maintained in the periphery at relatively constant levels via mechanisms that control expansion and contraction and are associated with competition for homeostatic cytokines. It has been shown that in a lymphopenic environment naive T cells undergo expansion due, at least in part, to additional availability of IL-7. We have previously found that T cell-intrinsic deletion of TNFR-associated factor (TRAF) 6 (TRAF6ΔT) in mice results in diminished peripheral CD8 T cell numbers. In this study, we report that whereas naive TRAF6ΔT CD8 T cells exhibit normal survival when transferred into a normal T cell pool, proliferation of naive TRAF6ΔT CD8 T cells under lymphopenic conditions is defective. We identified IL-18 as a TRAF6-activating factor capable of enhancing lymphopenia-induced proliferation (LIP) in vivo, and that IL-18 synergizes with high-dose IL-7 in a TRAF6-dependent manner to induce slow, LIP/homeostatic-like proliferation of naive CD8 T cells in vitro. IL-7 and IL-18 act synergistically to upregulate expression of IL-18R genes, thereby enhancing IL-18 activity. In this context, IL-18R signaling increases PI3K activation and was found to sensitize naive CD8 T cells to a model noncognate self-peptide ligand in a way that conventional costimulation via CD28 could not. We propose that synergistic sensitization by IL-7 and IL-18 to self-peptide ligand may represent a novel costimulatory pathway for LIP.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
7-Aminoactinomycin D, ~97% (HPLC), powder
Sigma-Aldrich
5(6)-Carboxyfluorescein diacetate N-succinimidyl ester, BioReagent, suitable for fluorescence, ≥90% (HPLC)
Sigma-Aldrich
5-Carboxy-fluorescein diacetate N-succinimidyl ester, suitable for fluorescence, ≥95.0% (HPLC)
Sigma-Aldrich
Sodium orthovanadate, ≥90% (titration)
Sigma-Aldrich
Sodium orthovanadate, 99.98% trace metals basis
Supelco
HEPES, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioUltra, Molecular Biology, ≥99.5% (T)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
SAFC
L-Glutamine
Supelco
L-Glutamine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Supelco
L-Glutamine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Glutamine
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O