Merck
  • Oligomerization of optineurin and its oxidative stress- or E50K mutation-driven covalent cross-linking: possible relationship with glaucoma pathology.
Oligomerization of optineurin and its oxidative stress- or E50K mutation-driven covalent cross-linking: possible relationship with glaucoma pathology.
PloS one (2014-07-02)
Jie Gao, Masafumi Ohtsubo, Yoshihiro Hotta, Shinsei Minoshima
ABSTRAKT
The optineurin gene, OPTN, is one of the causative genes of primary open-angle glaucoma. Although oligomerization of optineurin in cultured cells was previously observed by gel filtration analysis and blue native gel electrophoresis (BNE), little is known about the characteristics of optineurin oligomers. Here, we aimed to analyze the oligomeric state of optineurin and factors affecting oligomerization, such as environmental stimuli or mutations in OPTN. Using BNE or immunoprecipitation followed by sodium dodecyl sulfate-polyacrylamide gel electrophoresis (SDS-PAGE), we demonstrated that both endogenous and transfected optineurin exist as oligomers, rather than monomers, in NIH3T3 cells. We also applied an in situ proximity ligation assay to visualize the self-interaction of optineurin in fixed HeLaS3 cells and found that the optineurin oligomers were localized diffusely in the cytoplasm. Optineurin oligomers were usually detected as a single band of a size equal to that of the optineurin monomer upon SDS-PAGE, while an additional protein band of a larger size was observed when cells were treated with H2O2. We showed that larger protein complex is optineurin oligomers by immunoprecipitation and termed it covalent optineurin oligomers. In cells expressing OPTN bearing the most common glaucoma-associated mutation, E50K, covalent oligomers were formed even without H2O2 stimulation. Antioxidants inhibited the formation of E50K-induced covalent oligomers to various degrees. A series of truncated constructs of OPTN was used to reveal that covalent oligomers may be optineurin trimers and that the ubiquitin-binding domain is essential for formation of these trimers. Our results indicated that optineurin trimers may be the basic unit of these oligomers. The oligomeric state can be affected by many factors that induce covalent bonds, such as H2O2 or E50K, as demonstrated here; this provides novel insights into the pathogenicity of E50K. Furthermore, regulation of the oligomeric state should be studied in the future.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Ascorbic Acid, Pharmaceutical Secondary Standard; Certified Reference Material
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Anti-α-Tubulin antibody, Mouse monoclonal, clone AA13, purified from hybridoma cell culture
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
ANTI-FLAG® M2 antibody, Mouse monoclonal, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Informacje o cenach i dostępności nie są obecnie dostępne.
Ascorbic acid, European Pharmacopoeia (EP) Reference Standard
Informacje o cenach i dostępności nie są obecnie dostępne.
USP
Kwas askorbinowy, United States Pharmacopeia (USP) Reference Standard
Informacje o cenach i dostępności nie są obecnie dostępne.
Supelco
Hydrogen peroxide solution, 30 % (w/w), for ultratrace analysis
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
Informacje o cenach i dostępności nie są obecnie dostępne.
Supelco
L-Ascorbic acid, analytical standard
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
BIS-TRIS, BioPerformance Certified, suitable for cell culture, suitable for insect cell culture, ≥98.0%
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
BIS-TRIS, BioXtra, ≥98.0% (titration)
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
BIS-TRIS, ≥98.0% (titration)
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Informacje o cenach i dostępności nie są obecnie dostępne.
Supelco
Hydrogen peroxide solution, ≥30%, for trace analysis
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Hydrogen peroxide solution, tested according to Ph. Eur.
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
L-Ascorbic acid, 99%
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
BIS-TRIS, BioUltra, ≥99.0% (NT)
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
L-Ascorbic acid, tested according to Ph. Eur.
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
Informacje o cenach i dostępności nie są obecnie dostępne.
Millipore
Hydrogen peroxide solution, 3%, suitable for microbiology
Informacje o cenach i dostępności nie są obecnie dostępne.
Supelco
L-Ascorbic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
Informacje o cenach i dostępności nie są obecnie dostępne.