Merck
  • Validation of an automated solid-phase extraction method for the analysis of 23 opioids, cocaine, and metabolites in urine with ultra-performance liquid chromatography-tandem mass spectrometry.
Validation of an automated solid-phase extraction method for the analysis of 23 opioids, cocaine, and metabolites in urine with ultra-performance liquid chromatography-tandem mass spectrometry.
Journal of analytical toxicology (2014-05-03)
María del Mar Ramírez Fernández, Filip Van Durme, Sarah M R Wille, Vincent di Fazio, Natalie Kummer, Nele Samyn
ABSTRAKT
The aim of this work was to automate a sample preparation procedure extracting morphine, hydromorphone, oxymorphone, norcodeine, codeine, dihydrocodeine, oxycodone, 6-monoacetyl-morphine, hydrocodone, ethylmorphine, benzoylecgonine, cocaine, cocaethylene, tramadol, meperidine, pentazocine, fentanyl, norfentanyl, buprenorphine, norbuprenorphine, propoxyphene, methadone and 2-ethylidene-1,5-dimethyl-3,3-diphenylpyrrolidine from urine samples. Samples were extracted by solid-phase extraction (SPE) with cation exchange cartridges using a TECAN Freedom Evo 100 base robotic system, including a hydrolysis step previous extraction when required. Block modules were carefully selected in order to use the same consumable material as in manual procedures to reduce cost and/or manual sample transfers. Moreover, the present configuration included pressure monitoring pipetting increasing pipetting accuracy and detecting sampling errors. The compounds were then separated in a chromatographic run of 9 min using a BEH Phenyl analytical column on a ultra-performance liquid chromatography-tandem mass spectrometry system. Optimization of the SPE was performed with different wash conditions and elution solvents. Intra- and inter-day relative standard deviations (RSDs) were within ±15% and bias was within ±15% for most of the compounds. Recovery was >69% (RSD < 11%) and matrix effects ranged from 1 to 26% when compensated with the internal standard. The limits of quantification ranged from 3 to 25 ng/mL depending on the compound. No cross-contamination in the automated SPE system was observed. The extracted samples were stable for 72 h in the autosampler (4°C). This method was applied to authentic samples (from forensic and toxicology cases) and to proficiency testing schemes containing cocaine, heroin, buprenorphine and methadone, offering fast and reliable results. Automation resulted in improved precision and accuracy, and a minimum operator intervention, leading to safer sample handling and less time-consuming procedures.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Ammonium formate, ≥99.995% trace metals basis
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Ammonia solution, 0.4 M in THF
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Ammonia-14N, 99.99 atom % 14N
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Ammonia solution, 2.0 M in isopropanol
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Ammonia solution, 4 M in methanol
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Ammonium formate, reagent grade, 97%
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Ammonia solution, 2.0 M in ethanol
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Ammonia solution, 0.4 M in dioxane
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Ammonia solution, 7 N in methanol
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Ammonia, puriss., anhydrous, ≥99.95%
Informacje o cenach i dostępności nie są obecnie dostępne.
Supelco
Ammonium formate, eluent additive for LC-MS, LiChropur, ≥99.0%
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Ammonium formate, BioUltra, ≥99.0% (calc. based on dry substance, NT)
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Ammonium formate solution, BioUltra, 10 M in H2O
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Isopropyl alcohol, meets USP testing specifications
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Formic acid, reagent grade, ≥95%
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Formic acid, ACS reagent, ≥96%
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
2-Propanol, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
2-Propanol, suitable for HPLC, 99.9%
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Dichloromethane, ACS reagent, ≥99.5%, contains 40-150 ppm amylene as stabilizer
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Dichloromethane, biotech. grade, 99.9%, contains 40-150 ppm amylene as stabilizer
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
2-Propanol, suitable for HPLC, 99.5%
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Formic acid, ACS reagent, ≥88%
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Dichloromethane, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains 50-150 ppm amylene as stabilizer
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
2-Propanol, Laboratory Reagent, ≥99.5%
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Dichloromethane, suitable for HPLC, ≥99.9%, contains 40-150 ppm amylene as stabilizer
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Informacje o cenach i dostępności nie są obecnie dostępne.
Supelco
Methylene Chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Informacje o cenach i dostępności nie są obecnie dostępne.
Supelco
2-Propanol, Pharmaceutical Secondary Standard; Certified Reference Material
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
2-Propanol, electronic grade, 99.999% trace metals basis
Informacje o cenach i dostępności nie są obecnie dostępne.