Merck
Pedobacter bambusae sp. nov., isolated from soil of a bamboo plantation.
Antonie van Leeuwenhoek (2014-12-19)
Kyung Hwa Won, MooChang Kook, Tae-Hoo Yi
ABSTRAKT
An ivory white, rod-shaped, non-motile, non-spore forming, Gram-staining-negative, and aerobic bacterium designated THG-G118T, was isolated from a soil sample collected in a bamboo plantation in Seoul, Republic of Korea. The 16S rRNA gene sequence analysis revealed a clear affiliation of strain THG-G118T with the genus Pedobacter. Strain THG-G118T was found to show the closest phylogenetic relationship with Pedobacter metabolipauper WB2.3-71T with 16S rRNA gene sequence similarity of 97.3 %. The major cellular fatty acids of strain THG-G118T were identified as C16:0, C18:0, iso-C15:0 and summed feature 3 (C16:1 ω7c and/or C16:1 ω6c), and the predominant menaquinone as menaquinone MK-7. The major polar lipids were found to be phosphatidylethanolamine (PE), unidentified phospholipid and unidentified aminophospholipids. Strain THG-G118T was found to contain ceramide phosphorylethanolamine 2 (CerPE-2) and dihydrosphingosine as the major ceramide. The G+C content of the genomic DNA was found to be 41.7 mol%. DNA-DNA hybridization experiments revealed a low level of DNA-DNA relatedness (<42.5 %) between strain THG-G118T and its closest relatives. The phenotypic, chemotaxonomic and phylogenetic data showed that strain THG-G118T could be clearly distinguished from the members of the genus Pedobacter. Based on the results of a polyphasic study, a new species, Pedobacter bambusae sp. nov., is proposed. The type strain is THG-G118T (=KACC 17544T = JCM 19364T).
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
USP
1,4-benzochinon, United States Pharmacopeia (USP) Reference Standard
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Supelco
1,4-Benzoquinone, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Iron(III) citrate, technical grade
Supelco
Hydrogen chloride – methanol solution, ~1.25 m HCl (T), derivatization grade (GC derivatization), LiChropur
Supelco
Hydrogen chloride – 2-propanol solution, ~1.25 M HCl (T), derivatization grade (GC derivatization), LiChropur
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Supelco
Iron(III) citrate tribasic monohydrate, 18-20% Fe basis (T)
Sigma-Aldrich
Sodium chloride, BioUltra, Molecular Biology, ≥99.5% (AT)
Supelco
p-Benzoquinone, for spectrophotometric det. of amines, ≥99.5% (HPLC)
Sigma-Aldrich
p-Benzoquinone, reagent grade, ≥98%