Merck
  • Sialic acid-binding immunoglobulin-type lectin H-positive plasmacytoid dendritic cells drive spontaneous lupus-like disease development in B6.Nba2 mice.
Sialic acid-binding immunoglobulin-type lectin H-positive plasmacytoid dendritic cells drive spontaneous lupus-like disease development in B6.Nba2 mice.
Arthritis & rheumatology (Hoboken, N.J.) (2014-12-17)
Laura M Davison, Trine N Jørgensen
ABSTRAKT
Patients with systemic lupus erythematosus (SLE) often present with elevated levels of interferon-α (IFNα) in serum. Plasmacytoid dendritic cells (pDCs) have been suggested to be the primary source of IFNα in SLE due to their capacity to produce high levels of IFNα. During viral infection, a subset of pDCs expressing sialic acid-binding immunoglobulin-type lectin H (Siglec H) produces the majority of pDC-derived IFNα. The aim of this study was to provide evidence that Siglec H-positive pDCs are pathogenic in the IFNα-dependent B6.Nba2 mouse model of lupus. B6.Nba2 blood dendritic cell antigen 2 (BDCA-2)-diphtheria toxin receptor (DTR)-transgenic (Tg) mice were treated intraperitoneally with DT 3 times weekly starting at 4 weeks or 12 weeks of age and analyzed at 12 weeks and 18 weeks of age, respectively. Lupus-like disease development was measured by the presence of elevated levels of autoantibodies in serum (as determined by enzyme-linked immunosorbent assay), increased expression of IFN-inducible genes (as determined by real-time reverse transcription-polymerase chain reaction), increased IgG immune complex deposition in kidney glomeruli (as determined by immunofluorescence staining), spontaneous lymphocyte activation, and differentiation of B cells into antibody-producing plasma cells (as determined by flow cytometry). B6.Nba2 mice in which Siglec H-positive pDCs were depleted for 6-8 weeks displayed reduced levels of IFNα-induced gene transcripts and decreased anti-chromatin autoantibody levels in serum, and significantly fewer activated splenic T cells and B cells, germinal center B cells, follicular helper T cells, and splenic plasma cells. In 18-week-old mice, IgG immune complex deposition in kidney glomeruli was similarly reduced. The development of lupus-like disease in congenic B6.Nba2 mice depends on Siglec H-positive pDCs. We suggest that depletion of Siglec H-positive pDCs represents a novel cellular target in SLE.
MATERIAŁY
Numer produktu
Marka
Opis produktu
USP
Citric acid, United States Pharmacopeia (USP) Reference Standard
Citric acid, anhydrous, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
2-Hydroxybutyric acid sodium salt, 97%
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, ≥90% (HPLC)
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, BioReagent, suitable for fluorescence, mixture of 2 components, ≥90% (HPLC)
Sigma-Aldrich
Citric acid, anhydrous, suitable for cell culture, suitable for plant cell culture
Sigma-Aldrich
Citric acid, ACS reagent, ≥99.5%
Sigma-Aldrich
Citric acid, BioUltra, anhydrous, ≥99.5% (T)
Sigma-Aldrich
Citric acid, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.5%
Supelco
Citric acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Citric acid, Anhydrous, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Citric acid, 99%
Sigma-Aldrich
Citric acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Citric acid, meets analytical specification of Ph. Eur., BP, USP, E330, anhydrous, 99.5-100.5% (based on anhydrous substance)
Sigma-Aldrich
Pyridinium dichromate, 98%
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, suitable for protein labeling, ≥90% (HPLC), powder
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)