Merck
Expression and function of Ccbe1 in the chick early cardiogenic regions are required for correct heart development.
PloS one (2014-12-30)
João Furtado, Margaret Bento, Elizabeth Correia, José Manuel Inácio, José António Belo
ABSTRAKT
During the course of a differential screen to identify transcripts specific for chick heart/hemangioblast precursor cells, we have identified Ccbe1 (Collagen and calcium-binding EGF-like domain 1). While the importance of Ccbe1 for the development of the lymphatic system is now well demonstrated, its role in cardiac formation remained unknown. Here we show by whole-mount in situ hybridization analysis that cCcbe1 mRNA is initially detected in early cardiac progenitors of the two bilateral cardiogenic fields (HH4), and at later stages on the second heart field (HH9-18). Furthermore, cCcbe1 is expressed in multipotent and highly proliferative cardiac progenitors. We characterized the role of cCcbe1 during early cardiogenesis by performing functional studies. Upon morpholino-induced cCcbe1 knockdown, the chick embryos displayed heart malformations, which include aberrant fusion of the heart fields, leading to incomplete terminal differentiation of the cardiomyocytes. cCcbe1 overexpression also resulted in severe heart defects, including cardia bifida. Altogether, our data demonstrate that although cardiac progenitors cells are specified in cCcbe1 morphants, the migration and proliferation of cardiac precursors cells are impaired, suggesting that cCcbe1 is a key gene during early heart development.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Glycerin, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Gliceryna, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, BioXtra, ≥97 .0%
Supelco
HEPES, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, Molecular Biology, ≥97.0%
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, BioUltra, ≥99.0% (T)
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, ≥97.0%
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioUltra, Molecular Biology, ≥99.5% (T)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Glycerol, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
Glycerol, FCC, FG
Supelco
Glycerol, analytical standard
Sigma-Aldrich
Glycerin, meets USP testing specifications
Sigma-Aldrich
Glycerol, Molecular Biology, ≥99.0%
Sigma-Aldrich
Glycerol, BioUltra, Molecular Biology, anhydrous, ≥99.5% (GC)
Sigma-Aldrich
Glycerol, BioXtra, ≥99% (GC)
Sigma-Aldrich
Glycerol, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Glycerol, ≥99.5%
Sigma-Aldrich
Glycerol, tested according to Ph. Eur., anhydrous
Sigma-Aldrich
Glycerol, ACS reagent, ≥99.5%
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O