Merck
Expansion of specialized epidermis induced by hormonal state and mechanical strain.
Mechanisms of development (2015-02-15)
Hsin-Jung Wu, Teresa Easwaran, Carlos D Offutt, Richard Levi Elgar, Dan F Spandau, Sachiko Koyama, John Foley
ABSTRAKT
In mammals, some sites of specialized skin such as the palms, soles, and lips grow proportionally with the animal. However, other types of specialized skin such as the nipple and anal/genital region are dramatically altered with changes of reproductive status. The specific cell types that mediate the growth of these sites have not been identified. In the mouse, we observed a dramatic expansion of the specialized epidermis of the nipple, coupled to changes in connective tissue and hair shaft density, which we designate as areola formation. During this process thymidine analog uptake was elevated in the epidermis and hair follicles. Although there were no changes in connective tissue cell proliferation, we did observe an altered expression of extracellular matrix genes. In addition, the fibroblasts of the virgin nipple areola and region showed increased transcript and protein levels for estrogen, progesterone, relaxin, and oxytocin relative to those of ventral skin. To determine the role of pregnancy, lactation hormonal milieu, and localized mechanical strain on areola formation, we created models that separated these stimuli and evaluated changes in gross structure, proliferation and protein expression. While modest increases of epidermal proliferation and remodeling of connective tissue occurred as a result of individual stimuli, areola formation required exposure to pregnancy hormones, as well as mechanical strain.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Sodium phosphate dibasic solution, BioUltra, 0.5 M in H2O
Sigma-Aldrich
Formaldehyde-12C solution, 20% in H2O, 99.9 atom % 12C
Sigma-Aldrich
Picric acid solution, 1.3% in H2O (saturated)
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Supelco
Phosphate Standard for IC, TraceCERT®, 1000 mg/L phosphate in water (nominal concentration)
Supelco
Acetic acid, analytical standard
Sigma-Aldrich
Acetic acid, suitable for luminescence, BioUltra, ≥99.5% (GC)
Supelco
Picric acid solution, 100 μg/mL in acetonitrile, PESTANAL®, analytical standard
Sigma-Aldrich
Acetic acid, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Acetic acid, glacial, ≥99.99% trace metals basis
Sigma-Aldrich
Acetic acid, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Acetic acid solution, suitable for HPLC
Sigma-Aldrich
Przeciwciało anty-fosfo-CREB (Ser133), Upstate®, from rabbit
Supelco
Formaldehyde solution, stabilized with methanol, ~37 wt. % in H2O, certified reference material
Sigma-Aldrich
Progesterone, meets USP testing specifications
Sigma-Aldrich
Formaldehyde solution, tested according to Ph. Eur.
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Sigma-Aldrich
Formaldehyde solution, Molecular Biology, 36.5-38% in H2O
Sigma-Aldrich
Progesterone, powder, BioReagent, suitable for cell culture
Supelco
Progesterone, Pharmaceutical Secondary Standard; Certified Reference Material
Progesterone, European Pharmacopoeia (EP) Reference Standard
USP
Progesterone, United States Pharmacopeia (USP) Reference Standard
Progesterone for peak identification, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Progesterone, ≥99%
Sigma-Aldrich
Progesterone, γ-irradiated, BioXtra, suitable for cell culture
Supelco
Progesterone, VETRANAL®, analytical standard
Sigma-Aldrich
Formaldehyde solution, Molecular Biology, BioReagent, ≥36.0% in H2O (T)