Merck
PI3K signalling is required for a TGFβ-induced epithelial-mesenchymal-like transition (EMT-like) in human melanoma cells.
Experimental dermatology (2014-11-05)
Natalie C Schlegel, Anina von Planta, Daniel S Widmer, Reinhard Dummer, Gerhard Christofori
ABSTRAKT
Epithelial to mesenchymal transition (EMT) is a programme defined in epithelial cells and recognized as playing a critical role in cancer progression. Although melanoma is not a cancer of epithelial cells, hallmarks of EMT have been described to play a critical role in melanoma progression. Here, we demonstrate that long-term TGFβ exposure can induce a dedifferentiated EMT-like state resembling a previously described invasive phenotype (EMT-like). TGFβ-induced EMT-like is marked by the downregulation of melanocyte differentiation markers, such as MITF, and the upregulation of mesenchymal markers, such as N-cadherin, and an increase in melanoma cell migration and cell invasion. Pharmacological interference shows the dependency of TGFβ-induced EMT-like on the activation of the PDGF signalling pathway and the subsequent activation of PI3K in human melanoma cells. Together, the data provide novel insights into the transcriptional plasticity of melanoma cells that might contribute to tumor progression in patients and propose avenues to therapeutic interventions.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Supelco
Glycerin, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium pyruvate, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
USP
Gliceryna, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium pyruvate, BioXtra, ≥99%
Sigma-Aldrich
Sodium pyruvate, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium pyruvate, Hybri-Max, powder, suitable for hybridoma
Sigma-Aldrich
Anti-Fibronectin antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Sodium pyruvate, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, Molecular Biology, ~1 M in H2O
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
SB 431542 hydrate, ≥98% (HPLC), powder
Sigma-Aldrich
1,3-Dimethyl-2-imidazolidinone, absolute, over molecular sieve (H2O ≤0.04%), ≥99.5% (GC)
Sigma-Aldrich
1,3-Dimethyl-2-imidazolidinone, ≥99.0% (GC)
Sigma-Aldrich
1,3-Dimethyl-2-imidazolidinone, reagent grade
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, Molecular Biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Sodium Fluoride Solution
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC)