Merck
  • Human platelet lysate successfully promotes proliferation and subsequent chondrogenic differentiation of adipose-derived stem cells: a comparison with articular chondrocytes.
Human platelet lysate successfully promotes proliferation and subsequent chondrogenic differentiation of adipose-derived stem cells: a comparison with articular chondrocytes.
Journal of tissue engineering and regenerative medicine (2013-01-11)
F Hildner, M J Eder, K Hofer, J Aberl, H Redl, M van Griensven, C Gabriel, A Peterbauer-Scherb
ABSTRAKT
Fetal calf serum (FCS) bears a potential risk for carrying diseases and eliciting immune reactions. Nevertheless, it still represents the gold standard as medium supplement in cell culture. In the present study, human platelet lysate (PL) was tested as an alternative to FCS for the expansion and subsequent chondrogenic differentiation of human adipose-derived stem cells (ASCs). ASCs were expanded with 10% FCS (group F) or 5% PL (group P). Subsequently, three-dimensional (3D) micromass pellets were created and cultured for 5 weeks in chondrogenic differentiation medium. Additionally, the de- and redifferentiation potential of human articular chondrocytes (HACs) was evaluated and compared to ASCs. Both HACs and ASCs cultured with PL showed strongly enhanced proliferation rates. Redifferentiation of HACs was possible for cells expanded up to 3.3 population doublings (PD). At this stage, PL-expanded HACs demonstrated better redifferentiation potential than FCS-expanded cells. ASCs could also be differentiated following extended passaging. Glycosaminoglycan (GAG) quantification and qRT-PCR of 10 cartilage related markers demonstrated a tendency for increased chondrogenic differentiation of PL-expanded ASCs compared to cells expanded with FCS. Histologically, collagen type II but also collagen type X was mainly present in group P. The present study demonstrates that PL strongly induces proliferation of ASCs, while the chondrogenic differentiation potential is retained. HACs also showed enhanced proliferation and even better redifferentiation when previously expanded with PL. This suggests that PL is superior to FCS as a supplement for the expansion of ASCs and HACs, particularly with regard to chondrogenic (re)differentiation.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Selenium, foil, 25x25mm, thickness 3mm, 99.95%
Selenium, pellets, < 5mm, ≥99.999%
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
Sigma-Aldrich
Selenium, pellets, <5 mm particle size, ≥99.999% trace metals basis
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
Sigma-Aldrich
5-(N-Methyl-N-isobutyl)­amiloride, ≥98% (TLC), powder
Sigma-Aldrich
L-Cysteine hydrochloride, anhydrous, ≥98% (TLC)
Sigma-Aldrich
Retinoic acid, ≥98% (HPLC), powder
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
L-Cysteine hydrochloride, anhydrous, ≥99.0% (RT)
Sigma-Aldrich
L-Ascorbic acid, 99%
Sigma-Aldrich
Selenium, powder, −100 mesh, 99.99% trace metals basis
Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
Linoleic acid, technical, 58-74% (GC)
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
Linoleic acid, liquid, BioReagent, suitable for cell culture
Sigma-Aldrich
L-Cysteine hydrochloride, anhydrous, from non-animal source, BioReagent, suitable for cell culture, ≥98.0%
Sigma-Aldrich
Selenium, powder, −100 mesh, ≥99.5% trace metals basis
Sigma-Aldrich
Selenium, pellets, <5 mm, ≥99.99% trace metals basis
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
Sigma-Aldrich
Phenylacetic acid, ≥99%, FCC, FG
Sigma-Aldrich
Phenylacetic acid, 99%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Dexamethasone, ≥98% (HPLC), powder
Sigma-Aldrich
Dexamethasone, powder, γ-irradiated, BioXtra, suitable for cell culture, ≥80% (HPLC)