Merck
Multiplexed paper analytical device for quantification of metals using distance-based detection.
Lab on a chip (2015-05-27)
David M Cate, Scott D Noblitt, John Volckens, Charles S Henry
ABSTRAKT
Exposure to metal-containing aerosols has been linked with adverse health outcomes for almost every organ in the human body. Commercially available techniques for quantifying particulate metals are time-intensive, laborious, and expensive; often sample analysis exceeds $100. We report a simple technique, based upon a distance-based detection motif, for quantifying metal concentrations of Ni, Cu, and Fe in airborne particulate matter using microfluidic paper-based analytical devices. Paper substrates are used to create sensors that are self-contained, self-timing, and require only a drop of sample for operation. Unlike other colorimetric approaches in paper microfluidics that rely on optical instrumentation for analysis, with distance-based detection, analyte is quantified visually based on the distance of a colorimetric reaction, similar to reading temperature on a thermometer. To demonstrate the effectiveness of this approach, Ni, Cu, and Fe were measured individually in single-channel devices; detection limits as low as 0.1, 0.1, and 0.05 μg were reported for Ni, Cu, and Fe. Multiplexed analysis of all three metals was achieved with detection limits of 1, 5, and 1 μg for Ni, Cu, and Fe. We also extended the dynamic range for multi-analyte detection by printing concentration gradients of colorimetric reagents using an off-the-shelf inkjet printer. Analyte selectivity was demonstrated for common interferences. To demonstrate utility of the method, Ni, Cu, and Fe were measured from samples of certified welding fume; levels measured with paper sensors matched known values determined gravimetrically.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Sodium hydroxide, ultra dry, powder or crystals, 99.99% trace metals basis
Sigma-Aldrich
Trizma® base, BioUltra, Molecular Biology, ≥99.8% (T)
Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
Sodium acetate solution, BioUltra, Molecular Biology, ~3 M in H2O
Sigma-Aldrich
Trizma® base, ≥99.0% (T)
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, Molecular Biology, 10 M in H2O
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Sodium Acetate Anhydrous, >99%, FG
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
Tris(hydroxymethyl)aminomethane, ACS reagent, ≥99.8%
Sigma-Aldrich
Ammonium acetate, 99.999% trace metals basis
Sigma-Aldrich
L-Ascorbic acid, 99%
Sigma-Aldrich
Sodium acetate, 99.995% trace metals basis
Sigma-Aldrich
Acetic acid, suitable for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, Molecular Biology
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
Sigma-Aldrich
Sodium Fluoride Solution
Sigma-Aldrich
Trizma® base, ≥99.9% (titration), crystalline
Sigma-Aldrich
Ammonium acetate, BioXtra, ≥98%
Sigma-Aldrich
Ammonium acetate, Molecular Biology, ≥98%
Sigma-Aldrich
Ammonium acetate, reagent grade, ≥98%
Sigma-Aldrich
Ammonium acetate solution, Molecular Biology, 7.5 M