Merck
Heme oxygenase 1-mediated novel anti-inflammatory activities of Salvia plebeia and its active components.
Journal of ethnopharmacology (2015-09-01)
Muhammad Akram, Ahmed Shah Syed, Kyeong-A Kim, Jong Soo Lee, Sun-Young Chang, Chul Young Kim, Ok-Nam Bae
ABSTRAKT
Salvia plebeia R. Br. (SP) has been widely used as a traditional folk medicine for the treatment of infectious diseases and pain. An anti-inflammatory potential of SP has remains largely unknown. We tried to elucidate the principle mechanism and the active ingredients underlying the anti-inflammatory activities of SP. We investigated the protective activities of SP methanolic extract (SPME) and seven representative ingredients against inflammation. Quantitative analysis using HPLC-DAD-ESI/MS was conducted to determine the relative amounts of these seven active ingredients in SPME. Both in vitro murine macrophages and in vivo mouse models were employed to elucidate SP- and active ingredient-mediated anti-inflammatory effects. SPME significantly reduced inflammatory processes both in vivo in a TPA-induced ear edema model and in vitro in lipopolysaccharide (LPS)-activated macrophages. SPME decreased the release of nitric oxide (NO) and prostaglandin E2 (PGE2) and expression of inducible nitric oxide synthase (iNOS). Seven active components (luteoloside (C1), nepitrin (C2), homoplantagenin (C3), luteolin (C4), nepetin (C5), hispidulin (C6), and eupatorin (C7)) of SPME were analyzed and their relative concentrations were determined, demonstrating that C2, C3, C5 and C6 were present in higher amounts than were C1, C4, and C7. These major compounds inhibited NO and PGE2 production, and iNOS and COX-II protein expression through heme oxygenase-1 (HO-1) induction via activation of nuclear factor erythroid 2-related factor2 (Nrf2). Our data demonstrate that SPME possesses potent in vitro and in vivo anti-inflammatory activities. Nepetin and hispidulin, and their glycosides are the major active compounds in SPME, and their effects are mediated by Nrf2/HO-1 signaling. Taken together, we propose that SPME and its active ingredients may serve as novel therapeutic candidates for diseases associated with excessive inflammation.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Indomethacin, meets USP testing specifications
Sigma-Aldrich
2-Mercaptoethanol, ≥99.0%
Sigma-Aldrich
Indomethacin, 98.0-102.0%, meets EP testing specifications
Sigma-Aldrich
Prostaglandin E2, synthetic, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
Sigma-Aldrich
Prostaglandin E2, γ-irradiated, powder, BioXtra, suitable for cell culture
Sigma-Aldrich
Prostaglandin E2, ≥93% (HPLC), synthetic
Sigma-Aldrich
2-Mercaptoethanol, Molecular Biology, suitable for electrophoresis, suitable for cell culture, BioReagent, 99% (GC/titration)
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, ≥99% (TLC), film or powder
Sigma-Aldrich
PMA, for use in molecular biology applications, ≥99% (HPLC), Molecular Biology
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, synthetic, ≥98.0% (TLC)
Sigma-Aldrich
2-Mercaptoethanol, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Sodium nitrite, 99.999% trace metals basis
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Sodium nitrite, anhydrous, Redi-Dri, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Sigma-Aldrich
Sodium nitrite, ACS reagent, ≥97.0%
Sigma-Aldrich
Sodium nitrite, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%