Merck
Effective inactivation of Candida albicans biofilms by using supercritical carbon dioxide.
Bioprocess and biosystems engineering (2015-06-26)
Hyong Seok Park, Jungwoo Yang, Hee Jung Choi, Kyoung Heon Kim
ABSTRAKT
Present sterilization methods for biofilms in medical devices have limitations. Therefore, an alternative sterilization method using supercritical carbon dioxide (SC-CO2) was tested on Candida albicans biofilms. The effect of varying pressure, temperature, and treatment time on the inactivation of C. albicans spores in suspensions and in biofilms was examined. The parameters such as treatment time, pressure, and temperature that led to the complete inactivation of C. albicans biofilms ranged 5-20 min, 100-200 bar, and 35-45 °C, respectively. Notably, treatment of SC-CO2 at either 100 bar and 40 °C or 200 bar and 30 °C induced complete inactivation of spores within 5 min. Furthermore, it was found that wet biofilms (0.4 %, w/w) had higher sensitivity to SC-CO2 than dried biofilms. Finally, spore inactivation was confirmed by confocal laser scanning microscopy. In this study, the use of a low-temperature SC-CO2 sterilization method was proven to be effective in fungal biofilm inactivation, and the moisture content of biofilms was revealed to be the key factor for biofilm inactivation.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Sodium chloride solution, 0.85%
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Potassium chloride, ≥99.99% trace metals basis
Sigma-Aldrich
Potassium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Potassium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Potassium chloride solution, 0.075 M, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
Potassium chloride, 99.999% trace metals basis
Sigma-Aldrich
Potassium chloride solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Potassium chloride, BioXtra, ≥99.0%
Sigma-Aldrich
Potassium chloride, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Potassium chloride, Molecular Biology, ≥99.0%
Sigma-Aldrich
Sodium chloride, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Potassium chloride, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.0%
Sigma-Aldrich
Potassium phosphate monobasic, 99.99% trace metals basis