Merck
  • Analysis of sterigmatocystin in cereals, animal feed, seeds, beer and cheese by immunoaffinity column clean-up and HPLC and LC-MS/MS quantification.
Analysis of sterigmatocystin in cereals, animal feed, seeds, beer and cheese by immunoaffinity column clean-up and HPLC and LC-MS/MS quantification.
Food additives & contaminants. Part A, Chemistry, analysis, control, exposure & risk assessment (2015-10-31)
Elaine Marley, Phyllis Brown, Jennifer Mackie, Carol Donnelly, Joyce Wilcox, Amedeo Pietri, Susan Macdonald
ABSTRAKT
A method is reported for the analysis of sterigmatocystin in various food and feed matrices using a commercial sterigmatocystin immunoaffinity column (IAC) for sample clean-up prior to HPLC analysis by UV with mass spectrometric detection (LC-MS/MS). Cereals (wheat, oats, rye, maize and rice), sunflower seeds and animal feed were spiked with sterigmatocystin at levels from 0.75 to 50 µg kg(-1) to establish method performance. Using acetonitrile/water extraction followed by IAC clean-up, and analysis by HPLC with detection at 325 nm, recoveries ranged from 68% to 106%, with repeatability from 4.2% to 17.5%. The limit of quantification with UV detection in these matrices was 1.5 µg kg(-1). For the analysis of beer and cheese the sample preparation prior to IAC clean-up was changed to accommodate the different properties of the matrix, prior to analysis by LC-MS/MS. For beer and cheese spiked at 5.0 µg kg(-1) the recoveries were 94% and 104%, and precision (RSDs) were 1.9% and 2.9% respectively. The limits of quantification by LC-MS/MS in beer and cheese were 0.02 and 0.6 µg kg(-1) respectively. The sterigmatocystin IAC was demonstrated to provide an efficient clean-up of various matrices to enable this mycotoxin to be determined by either HPLC with UV detection or LC-MS/MS.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sterigmatocystin, powder, ≥98% (TLC)
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Ammonium formate, reagent grade, 97%
Sigma-Aldrich
Ammonium formate, ≥99.995% trace metals basis
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Sodium chloride, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%