Merck
Proteomic Profiling of Paclitaxel Treated Cells Identifies a Novel Mechanism of Drug Resistance Mediated by PDCD4.
Journal of proteome research (2015-05-01)
Hui Xu, Noah Dephoure, Huiying Sun, Haiyuan Zhang, Fangfang Fan, Jiawei Liu, Xuelian Ning, Shaochun Dai, Baogang Liu, Min Gao, Songbin Fu, Steven P Gygi, Chunshui Zhou
ABSTRAKT
Paclitaxel (PTX) is a widely used chemotherapeutic drug effective against numerous cancers. To elucidate cellular pathways targeted by PTX and identify novel mechanisms of PTX resistance, we used a SILAC based quantitative proteomic approach to evaluate global changes of cellular protein abundance in HeLa cells. We identified 347 proteins involved in a number of biological processes including spindle assembly, mitotic exit, and extracellular adhesion whose abundance changes upon PTX treatment. Notably, the tumor suppressor PDCD4 involved in translation suppression was down-regulated by PTX. We demonstrated that PDCD4 is a cell-cycle regulated protein and that changes in its abundance are sufficient to alter PTX sensitivity in multiple human cancer cell lines. Immunoprecipitation of PDCD4-RNA complexes and RT-PCR revealed that PDCD4 mediated PTX sensitivity acts through its interaction with mRNA of UBE2S, a ubiquitin K11 linkage conjugating enzyme critical for mitotic exit. Lastly, high levels of PDCD4 in lung cancer tissues are positively correlated with the longer overall survival time of the examined lung cancer patients with PTX involved adjuvant therapy. Therefore, our proteomic screen for paclitaxel targets not only provided novel insight into the cellular resistance to paclitaxel via the PDCD4-mitotic exit regulation axis, but also offered a predictive biomarker for paclitaxel-based personalized chemotherapy in the treatment of lung cancer.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
Sigma-Aldrich
Nocodazole, ≥99% (TLC), powder
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, Molecular Biology, ~1 M in H2O
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
L-Lysine, ≥98% (TLC)
Sigma-Aldrich
L-Lysine, crystallized, ≥98.0% (NT)
Supelco
DL-Dithiothreitol solution, 1 M in H2O
Sigma-Aldrich
Sodium chloride, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
SAFC
L-Arginine
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
L-Arginine, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Arginine, BioUltra, ≥99.5% (NT)