Merck
Estrogen modulation properties of mangiferin and quercetin and the mangiferin metabolite norathyriol.
Food & function (2015-05-06)
Ashley S Wilkinson, Meng-Wong Taing, Jean Thomas Pierson, Chun-Nam Lin, Ralf G Dietzgen, P Nicholas Shaw, Michael J Gidley, Gregory R Monteith, Sarah J Roberts-Thomson
ABSTRAKT
Mango fruit contain many bioactive compounds, some of which are transcription factor regulators. Estrogen receptor alpha (ERα) and beta (ERβ) are two regulators of gene transcription that are important in a variety of physiological processes and also in diseases including breast cancer. We examined the ability of the mango constituents quercetin, mangiferin, and the aglycone form of mangiferin, norathyriol, to activate both isoforms of the estrogen receptor. Quercetin and norathyriol decreased the viability of MCF-7 breast cancer cells whereas mangiferin had no effect on MCF-7 cells. We also determined that quercetin and mangiferin selectively activated ERα whereas norathyriol activated both ERα and ERβ. Despite quercetin, mangiferin and norathyriol having similar polyphenolic structural motifs, only norathyriol activated ERβ, showing that bioactive agents in mangoes have very specific biological effects. Such specificity may be important given the often-opposing roles of ERα and ERβ in breast cancer proliferation and other cellular processes.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, ~0.025 M in H2O
Sigma-Aldrich
Magnesium chloride solution, PCR Reagent, 25 mM MgCI2 solution for PCR
Sigma-Aldrich
Magnesium chloride solution, 0.1 M
Sigma-Aldrich
Magnesium chloride solution, Molecular Biology, 1.00 M±0.01 M
Sigma-Aldrich
Mangiferin Mangifera indica
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, 2 M in H2O
Sigma-Aldrich
Quercetin, ≥95% (HPLC), solid
Sigma-Aldrich
2-Mercaptoethanol, Molecular Biology, suitable for electrophoresis, suitable for cell culture, BioReagent, 99% (GC/titration)
Sigma-Aldrich
2-Mercaptoethanol, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
2-Mercaptoethanol, ≥99.0%
Sigma-Aldrich
GW9662, >98% (HPLC)
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Magnesium chloride, powder, <200 μm
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Sigma-Aldrich
Sodium phosphate, 96%
Sigma-Aldrich
Magnesium chloride, suitable for insect cell culture, BioReagent, ≥97.0%
Sigma-Aldrich
Magnesium chloride, ≥98%
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology