Merck
Igs as Substrates for Transglutaminase 2: Implications for Autoantibody Production in Celiac Disease.
Journal of immunology (Baltimore, Md. : 1950) (2015-10-28)
Rasmus Iversen, M Fleur du Pré, Roberto Di Niro, Ludvig M Sollid
ABSTRAKT
Autoantibodies specific for the enzyme transglutaminase 2 (TG2) are a hallmark of the gluten-sensitive enteropathy celiac disease. Production of the Abs is strictly dependent on exposure to dietary gluten proteins, thus raising the question how a foreign Ag (gluten) can induce an autoimmune response. It has been suggested that TG2-reactive B cells are activated by gluten-reactive T cells following receptor-mediated uptake of TG2-gluten complexes. In this study, we propose a revised model that is based on the ability of the BCR to serve as a substrate to TG2 and become cross-linked to gluten-derived peptides. We show that TG2-specific IgD molecules are preferred in the reaction and that binding of TG2 via a common epitope targeted by cells using the IgH variable gene segment (IGHV)5-51 results in more efficient cross-linking. Based on these findings we hypothesize that IgD-expressing B cells using IGHV5-51 are preferentially activated, and we suggest that this property can explain the previously reported low number of somatic mutations as well as the overrepresentation of IGHV5-51 among TG2-specific plasma cells in the celiac lesion. The model also couples gluten peptide uptake by TG2-reactive B cells directly to peptide deamidation, which is necessary for the activation of gluten-reactive T cells. It thereby provides a link between gluten deamidation, T cell activation, and the production of TG2-specific Abs. These are all key events in the development of celiac disease, and by connecting them the model may explain why the same enzyme that catalyzes gluten deamidation is also an autoantigen, something that is hardly coincidental.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Calcium chloride, powder, 99.99% trace metals basis
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.99% trace metals basis
Sigma-Aldrich
Calcium chloride solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Calcium chloride, anhydrous, BioReagent, suitable for insect cell culture, suitable for plant cell culture, ≥96.0%
Sigma-Aldrich
Calcium chloride
Sigma-Aldrich
Thrombopoietin human, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture, ≥98% (SDS-PAGE and HPLC)
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Thrombopoietin from mouse, recombinant, expressed in NSO cells, lyophilized powder, suitable for cell culture, >97% (SDS-PAGE)
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture