Merck
Anomalous behavior of visible light active TiO2 for the photocatalytic degradation of different Reactive dyes.
Photochemical & photobiological sciences : Official journal of the European Photochemistry Association and the European Society for Photobiology (2015-08-26)
Navneet Kaur, Satwant Kaur Shahi, Vasundhara Singh
ABSTRAKT
Nanocrystalline undoped, N-doped, N and metal codoped titania with different particle size, surface area, anatase phase content, crystallinity, band gap and zeta potential were synthesized by the sol-gel method. The photocatalytic activities of the synthesized TiO2 powders were compared by employing four different Reactive dyes. The order of photocatalytic activity observed for Reactive Red 198 dye (RR 198) was undoped = N,Cu codoped = N-doped > N,Fe codoped TiO2, Reactive Blue 4 dye (RB 4) was N,Cu codoped > N,Fe codoped > N-doped > undoped TiO2, Reactive Black 5 dye (RB 5) was N,Cu codoped* > undoped > N-doped > N,Fe codoped TiO2 and negligible degradation was observed for Reactive Orange 16 dye (RO 16). In this paper, the anomalous trend of the photocatalytic activity of various photocatalysts for the degradation of a particular class of dyes has been observed and accounted for based upon three parameters: mechanism of degradation, physicochemical properties of the catalyst and adsorption behavior based on the zeta potential. It was concluded that apart from these parameters, the substrate-specificity of the catalyst is also of equal importance in developing new catalysts for the photodegradation of dyes present in textile effluents.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Urea solution, 40 % (w/v) in H2O
Sigma-Aldrich
Urea-12C, 99.9 atom % 12C
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Acetic acid, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Acetic acid, suitable for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Acetic acid, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Acetic acid solution, suitable for HPLC
Sigma-Aldrich
Acetic acid, glacial, ≥99.99% trace metals basis
Sigma-Aldrich
Titanium(IV) butoxide, reagent grade, 97%
Sigma-Aldrich
Urea solution, BioUltra, ~8 M in H2O
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, purum, ≥99.0% (TLC)
Supelco
Urea, 8 M (after reconstitution with 16 mL high purity water)
Sigma-Aldrich
Urea, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99%
Sigma-Aldrich
Urea, ACS reagent, 99.0-100.5%
Sigma-Aldrich
Urea, BioUltra, Molecular Biology, 99% (T)
Sigma-Aldrich
Urea, meets USP testing specifications
Sigma-Aldrich
Urea, BioXtra, pH 7.5-9.5 (20 °C, 5 M in H2O)
Sigma-Aldrich
Urea, ReagentPlus®, ≥99.5%, pellets
Sigma-Aldrich
Urea, suitable for electrophoresis
Sigma-Aldrich
Urea, powder, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Urea, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%, 99.0-101.0% (calc. on dry substance)
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, ≥97.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%