Merck
Patient-derived hepatitis C virus inhibits CD4⁺ but not CD8⁺ T lymphocyte proliferation in primary T cells.
Virology journal (2015-06-19)
Sonya A MacParland, Annie Y Chen, Christopher P Corkum, Tram N Q Pham, Tomasz I Michalak
ABSTRAKT
Hepatitis C virus (HCV) can replicate in cells of the immune system and productively propagate in primary T lymphocytes in vitro. We aimed to determine whether exposure to authentic, patient-derived HCV can modify the proliferation capacity, susceptibility to apoptosis and phenotype of T cells. Primary total T cells from a healthy donor were used as targets and plasma-derived HCV from patients with chronic hepatitis C served as inocula. T cell phenotype was determined prior to and at different time points after exposure to HCV. T cell proliferation and apoptosis were measured by flow cytometry-based assays. The HCV inocula that induced the highest intracellular expression of HCV also caused a greatest shift in the T cell phenotype from predominantly CD4-positive to CD8-positive. This shift was associated with inhibition of CD4+ but not CD8+ T cell proliferation and did not coincide with altered apoptotic death of either cell subset. The data obtained imply that exposure to native HCV can have an impact on the relative frequencies of CD4+ and CD8+ T cells by selectively suppressing CD4(+) T lymphocyte proliferation and this may occur in both the presence and the absence of measurable HCV replication in these cells. If the virus exerts a similar effect in vivo, it may contribute to the impairment of virus-specific T cell response by altering cooperation between immune cell subsets.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
5(6)-Carboxyfluorescein diacetate N-succinimidyl ester, BioReagent, suitable for fluorescence, ≥90% (HPLC)
Sigma-Aldrich
5-Carboxy-fluorescein diacetate N-succinimidyl ester, suitable for fluorescence, ≥95.0% (HPLC)
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioUltra, Molecular Biology, ≥99.5% (T)
Sigma-Aldrich
Aphidicolin from Nigrospora sphaerica, ≥98% (HPLC), powder
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Sigma-Aldrich
6-Phosphonohexanoic acid, 97%
Sigma-Aldrich
Calcium chloride, powder, 99.99% trace metals basis
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.99% trace metals basis
Sigma-Aldrich
Calcium chloride, anhydrous, BioReagent, suitable for insect cell culture, suitable for plant cell culture, ≥96.0%
Sigma-Aldrich
Calcium chloride
Sigma-Aldrich
Calcium chloride solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O